Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26405471-26405744 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A1
Entrez Gene ID
11119
Gene Name
butyrophilin subfamily 3 member A1
Gene Aliases
BT3.1, BTF5, BTN3.1, CD277
UniGene
Hs.191510
Gene Chromosome Location
Chr.6: 26402237-26415216 on Build GRCh38
Transcripts
NM_001145008.1 NM_001145009.1 NM_007048.5 NM_194441.2 XM_005248833.2 XM_005248834.3 XM_006714983.2 XM_011514268.2

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NM_001145008.1 NM_001145009.1 NM_007048.5 NM_194441.2 XM_005248833.2 XM_005248834.3 XM_006714983.2 XM_011514268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7159142 c.85+2T>A p.? Chr.6: 26405650-26405650 on Build GRCh38
3860986 c.82T>C p.S28P Chr.6: 26405645-26405645 on Build GRCh38
5668867 c.13A>G p.S5G Chr.6: 26405576-26405576 on Build GRCh38
3860988 c.85+2T>C p.? Chr.6: 26405650-26405650 on Build GRCh38
7159140 c.85+2T>A p.? Chr.6: 26405650-26405650 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775494933 Chr.6: 26405649-26405649 on Build GRCh38
rs566411691 Chr.6: 26405681-26405681 on Build GRCh38
rs987410426 Chr.6: 26405555-26405555 on Build GRCh38
rs746012673 Chr.6: 26405629-26405629 on Build GRCh38
rs532906057 Chr.6: 26405531-26405531 on Build GRCh38
rs769191942 Chr.6: 26405654-26405654 on Build GRCh38
rs55676749 Chr.6: 26405588-26405588 on Build GRCh38
rs781510381 Chr.6: 26405620-26405620 on Build GRCh38
rs975118760 Chr.6: 26405596-26405596 on Build GRCh38
rs765193555 Chr.6: 26405535-26405535 on Build GRCh38
rs192573594 Chr.6: 26405672-26405672 on Build GRCh38
rs374569079 Chr.6: 26405684-26405684 on Build GRCh38
rs753390954 Chr.6: 26405601-26405601 on Build GRCh38
rs762189203 Chr.6: 26405665-26405665 on Build GRCh38
rs770798335 Chr.6: 26405613-26405613 on Build GRCh38
rs1011558552 Chr.6: 26405515-26405515 on Build GRCh38
rs963333249 Chr.6: 26405503-26405503 on Build GRCh38
rs764572510 Chr.6: 26405558-26405558 on Build GRCh38
rs1020956569 Chr.6: 26405527-26405527 on Build GRCh38
rs56161420 Chr.6: 26405607-26405607 on Build GRCh38
rs754288032 Chr.6: 26405526-26405527 on Build GRCh38
rs769935296 Chr.6: 26405646-26405646 on Build GRCh38
rs757923597 Chr.6: 26405638-26405644 on Build GRCh38
rs776073206 Chr.6: 26405554-26405554 on Build GRCh38
rs904782252 Chr.6: 26405648-26405648 on Build GRCh38
rs371938458 Chr.6: 26405676-26405676 on Build GRCh38
rs766105293 Chr.6: 26405600-26405600 on Build GRCh38
rs764935664 Chr.6: 26405685-26405685 on Build GRCh38
rs752290795 Chr.6: 26405697-26405697 on Build GRCh38
rs776692073 Chr.6: 26405532-26405532 on Build GRCh38
rs540333878 Chr.6: 26405597-26405597 on Build GRCh38
rs1001601096 Chr.6: 26405504-26405504 on Build GRCh38
rs973099425 Chr.6: 26405524-26405524 on Build GRCh38
rs369728603 Chr.6: 26405560-26405560 on Build GRCh38
rs878958419 Chr.6: 26405721-26405721 on Build GRCh38
rs746846979 Chr.6: 26405609-26405609 on Build GRCh38
rs746791964 Chr.6: 26405513-26405513 on Build GRCh38
rs535315381 Chr.6: 26405718-26405718 on Build GRCh38
rs763016689 Chr.6: 26405651-26405651 on Build GRCh38
rs774819205 Chr.6: 26405657-26405657 on Build GRCh38
rs893184829 Chr.6: 26405717-26405717 on Build GRCh38
rs374921833 Chr.6: 26405606-26405606 on Build GRCh38
rs1002006751 Chr.6: 26405683-26405683 on Build GRCh38
rs1034377325 Chr.6: 26405510-26405510 on Build GRCh38
rs776107351 Chr.6: 26405528-26405528 on Build GRCh38
rs753542420 Chr.6: 26405680-26405680 on Build GRCh38

View Full Product Details