Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 8914729-8915000 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKIP1
Entrez Gene ID
56672
Gene Name
A-kinase interacting protein 1
Gene Aliases
BCA3, C11orf17
UniGene
Hs.131180
Gene Chromosome Location
Chr.11: 8911117-8920079 on Build GRCh38
Transcripts
NM_001206646.1 NM_001206647.1 NM_001206648.1 NM_020642.3 NR_045417.1 NR_045418.1 XM_017018011.1 XM_017018012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7352946 c.395T>C p.I132T Chr.11: 8914917-8914917 on Build GRCh38
1246377 c.390_407del18 p.D131_Q136delDIGNGQ Chr.11: 8914912-8914929 on Build GRCh38
7355094 c.338C>A p.A113E Chr.11: 8914860-8914860 on Build GRCh38
7296391 c.371C>T p.S124L Chr.11: 8914893-8914893 on Build GRCh38
6223004 c.366C>T p.G122G Chr.11: 8914888-8914888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751144405 Chr.11: 8914911-8914911 on Build GRCh38
rs776599356 Chr.11: 8914893-8914893 on Build GRCh38
rs372405920 Chr.11: 8914957-8914957 on Build GRCh38
rs762849274 Chr.11: 8914879-8914879 on Build GRCh38
rs761865447 Chr.11: 8914934-8914935 on Build GRCh38
rs746977984 Chr.11: 8914772-8914772 on Build GRCh38
rs777816809 Chr.11: 8914931-8914931 on Build GRCh38
rs907842920 Chr.11: 8914789-8914789 on Build GRCh38
rs777259434 Chr.11: 8914964-8914964 on Build GRCh38
rs762995502 Chr.11: 8914807-8914807 on Build GRCh38
rs1049319322 Chr.11: 8914881-8914881 on Build GRCh38
rs985194588 Chr.11: 8914764-8914764 on Build GRCh38
rs757695894 Chr.11: 8914905-8914905 on Build GRCh38
rs768128986 Chr.11: 8914794-8914794 on Build GRCh38
rs754127774 Chr.11: 8914898-8914898 on Build GRCh38
rs746034338 Chr.11: 8914940-8914940 on Build GRCh38
rs927358670 Chr.11: 8914752-8914752 on Build GRCh38
rs372286619 Chr.11: 8914934-8914934 on Build GRCh38
rs754256215 Chr.11: 8914854-8914854 on Build GRCh38
rs759739590 Chr.11: 8914793-8914793 on Build GRCh38
rs780630443 Chr.11: 8914916-8914916 on Build GRCh38
rs183724545 Chr.11: 8914875-8914875 on Build GRCh38
rs186947057 Chr.11: 8914972-8914972 on Build GRCh38
rs951974279 Chr.11: 8914902-8914902 on Build GRCh38
rs376791560 Chr.11: 8914818-8914818 on Build GRCh38
rs753328281 Chr.11: 8914817-8914817 on Build GRCh38
rs762230416 Chr.11: 8914971-8914971 on Build GRCh38
rs78554101 Chr.11: 8914873-8914873 on Build GRCh38
rs775629675 Chr.11: 8914786-8914787 on Build GRCh38
rs772842815 Chr.11: 8914878-8914878 on Build GRCh38
rs79029701 Chr.11: 8914913-8914913 on Build GRCh38
rs530376117 Chr.11: 8914891-8914891 on Build GRCh38
rs889524655 Chr.11: 8914841-8914841 on Build GRCh38
rs770828800 Chr.11: 8914933-8914933 on Build GRCh38
rs200350441 Chr.11: 8914942-8914942 on Build GRCh38
rs777945419 Chr.11: 8914847-8914847 on Build GRCh38
rs767364125 Chr.11: 8914956-8914957 on Build GRCh38
rs35131475 Chr.11: 8914917-8914917 on Build GRCh38
rs564656115 Chr.11: 8914774-8914774 on Build GRCh38
rs764697362 Chr.11: 8914894-8914894 on Build GRCh38
rs751562462 Chr.11: 8914790-8914790 on Build GRCh38
rs141384258 Chr.11: 8914887-8914887 on Build GRCh38
rs567188870 Chr.11: 8914889-8914889 on Build GRCh38
rs764492452 Chr.11: 8914961-8914961 on Build GRCh38
rs73410030 Chr.11: 8914960-8914960 on Build GRCh38
rs144965150 Chr.11: 8914760-8914760 on Build GRCh38
rs1040574576 Chr.11: 8914799-8914799 on Build GRCh38
rs759687324 Chr.11: 8914888-8914888 on Build GRCh38
rs368030888 Chr.11: 8914864-8914864 on Build GRCh38
rs751503930 Chr.11: 8914932-8914933 on Build GRCh38
rs1050643201 Chr.11: 8914835-8914835 on Build GRCh38
rs746506425 Chr.11: 8914866-8914866 on Build GRCh38
rs779567750 Chr.11: 8914909-8914909 on Build GRCh38
rs867229313 Chr.11: 8914837-8914837 on Build GRCh38
rs771038583 Chr.11: 8914883-8914883 on Build GRCh38
rs764094759 Chr.11: 8914829-8914831 on Build GRCh38
rs771033387 Chr.11: 8914874-8914874 on Build GRCh38

View Full Product Details