Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38749795-38750054 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR5
Entrez Gene ID
79913
Gene Name
ARP5 actin-related protein 5 homolog
Gene Aliases
Arp5, INO80M
UniGene
Hs.371585
Gene Chromosome Location
Chr.20: 38748454-38772446 on Build GRCh38
Transcripts
NM_024855.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1411729 c.385G>A p.D129N Chr.20: 38750019-38750019 on Build GRCh38
4385141 c.410C>T p.A137V Chr.20: 38750044-38750044 on Build GRCh38
6593123 c.391C>T p.P131S Chr.20: 38750025-38750025 on Build GRCh38
4983478 c.406G>C p.E136Q Chr.20: 38750040-38750040 on Build GRCh38
6387369 c.392C>A p.P131H Chr.20: 38750026-38750026 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745689083 Chr.20: 38750012-38750012 on Build GRCh38
rs376328133 Chr.20: 38750025-38750025 on Build GRCh38
rs779741103 Chr.20: 38749891-38749891 on Build GRCh38
rs531752105 Chr.20: 38750014-38750014 on Build GRCh38
rs542978478 Chr.20: 38749928-38749928 on Build GRCh38
rs760643640 Chr.20: 38750008-38750008 on Build GRCh38
rs773997822 Chr.20: 38749976-38749976 on Build GRCh38
rs750701986 Chr.20: 38750020-38750020 on Build GRCh38
rs369906301 Chr.20: 38750028-38750028 on Build GRCh38
rs140420007 Chr.20: 38750031-38750031 on Build GRCh38
rs971791098 Chr.20: 38749941-38749941 on Build GRCh38
rs776643717 Chr.20: 38749914-38749914 on Build GRCh38
rs769386461 Chr.20: 38749878-38749878 on Build GRCh38
rs1051513921 Chr.20: 38749840-38749840 on Build GRCh38
rs899095132 Chr.20: 38749990-38749990 on Build GRCh38
rs748819877 Chr.20: 38749959-38749959 on Build GRCh38
rs1004654563 Chr.20: 38749867-38749867 on Build GRCh38
rs562841865 Chr.20: 38750003-38750003 on Build GRCh38
rs770292481 Chr.20: 38749965-38749965 on Build GRCh38
rs372446166 Chr.20: 38749988-38749988 on Build GRCh38
rs200852027 Chr.20: 38750030-38750030 on Build GRCh38
rs753920316 Chr.20: 38750009-38750009 on Build GRCh38
rs2254245 Chr.20: 38749865-38749865 on Build GRCh38
rs573975067 Chr.20: 38749913-38749913 on Build GRCh38
rs961737264 Chr.20: 38749930-38749930 on Build GRCh38
rs759149486 Chr.20: 38749978-38749978 on Build GRCh38

View Full Product Details