Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 83048354-83048621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNTL1
Entrez Gene ID
146712
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1
Gene Aliases
3-Gn-T8, B3GNT8, BGnT-8, beta-1, beta3Gn-T8, beta3GnTL1
UniGene
Hs.607824
Gene Chromosome Location
Chr.17: 82942149-83051845 on Build GRCh38
Transcripts
NM_001009905.2 NM_001320742.1 NR_135465.1 XM_006722272.3 XM_006722274.2 XM_011523532.2 XM_017024205.1 XM_017024207.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1184325 c.268C>A p.P90T Chr.17: 83048480-83048480 on Build GRCh38
986214 c.255C>T p.H85H Chr.17: 83048493-83048493 on Build GRCh38
1660719 c.266C>T p.S89F Chr.17: 83048482-83048482 on Build GRCh38
1710921 c.263C>T p.P88L Chr.17: 83048485-83048485 on Build GRCh38
5762789 c.256G>T p.D86Y Chr.17: 83048492-83048492 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758198080 Chr.17: 83048567-83048567 on Build GRCh38
rs139235297 Chr.17: 83048541-83048541 on Build GRCh38
rs753060519 Chr.17: 83048443-83048443 on Build GRCh38
rs10852789 Chr.17: 83048410-83048410 on Build GRCh38
rs374255430 Chr.17: 83048510-83048510 on Build GRCh38
rs866687785 Chr.17: 83048488-83048488 on Build GRCh38
rs927897234 Chr.17: 83048412-83048412 on Build GRCh38
rs372146084 Chr.17: 83048419-83048419 on Build GRCh38
rs776029794 Chr.17: 83048506-83048506 on Build GRCh38
rs777721301 Chr.17: 83048568-83048568 on Build GRCh38
rs778100186 Chr.17: 83048493-83048493 on Build GRCh38
rs779060909 Chr.17: 83048483-83048483 on Build GRCh38
rs763344445 Chr.17: 83048457-83048457 on Build GRCh38
rs200190461 Chr.17: 83048504-83048504 on Build GRCh38
rs757493255 Chr.17: 83048583-83048583 on Build GRCh38
rs781537874 Chr.17: 83048496-83048496 on Build GRCh38
rs373293017 Chr.17: 83048441-83048441 on Build GRCh38
rs932164843 Chr.17: 83048508-83048508 on Build GRCh38
rs376044870 Chr.17: 83048551-83048551 on Build GRCh38
rs370362164 Chr.17: 83048590-83048590 on Build GRCh38
rs776756158 Chr.17: 83048500-83048500 on Build GRCh38
rs75432825 Chr.17: 83048414-83048414 on Build GRCh38
rs755556988 Chr.17: 83048495-83048496 on Build GRCh38
rs760104555 Chr.17: 83048435-83048435 on Build GRCh38
rs760390547 Chr.17: 83048542-83048542 on Build GRCh38
rs752711722 Chr.17: 83048561-83048561 on Build GRCh38
rs181626136 Chr.17: 83048499-83048499 on Build GRCh38
rs754487338 Chr.17: 83048555-83048555 on Build GRCh38
rs755537365 Chr.17: 83048471-83048471 on Build GRCh38
rs753662161 Chr.17: 83048474-83048474 on Build GRCh38
rs746834338 Chr.17: 83048578-83048578 on Build GRCh38
rs377014413 Chr.17: 83048520-83048520 on Build GRCh38
rs745942236 Chr.17: 83048593-83048593 on Build GRCh38
rs772014583 Chr.17: 83048487-83048487 on Build GRCh38
rs143030444 Chr.17: 83048472-83048472 on Build GRCh38
rs771076029 Chr.17: 83048423-83048423 on Build GRCh38
rs1143006 Chr.17: 83048511-83048511 on Build GRCh38
rs377493899 Chr.17: 83048485-83048485 on Build GRCh38
rs754930933 Chr.17: 83048479-83048479 on Build GRCh38
rs772675170 Chr.17: 83048525-83048525 on Build GRCh38
rs747257029 Chr.17: 83048496-83048496 on Build GRCh38
rs553547179 Chr.17: 83048559-83048559 on Build GRCh38
rs202195967 Chr.17: 83048503-83048503 on Build GRCh38
rs766006076 Chr.17: 83048546-83048546 on Build GRCh38
rs750094617 Chr.17: 83048470-83048470 on Build GRCh38
rs143961054 Chr.17: 83048434-83048434 on Build GRCh38
rs767130690 Chr.17: 83048463-83048463 on Build GRCh38
rs533898331 Chr.17: 83048387-83048387 on Build GRCh38
rs983492277 Chr.17: 83048417-83048418 on Build GRCh38

View Full Product Details