Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 20647907-20648148 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB5
Entrez Gene ID
340273
Gene Name
ATP binding cassette subfamily B member 5
Gene Aliases
ABCB5alpha, ABCB5beta, EST422562
UniGene
Hs.404102
Gene Chromosome Location
Chr.7: 20615622-20757014 on Build GRCh38
Transcripts
NM_001163941.1 NM_001163942.1 NM_001163993.2 NM_178559.5 XM_011515367.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3637187 c.1186C>T p.P396S Chr.7: 20648058-20648058 on Build GRCh38
1488456 c.1190C>T p.S397L Chr.7: 20648062-20648062 on Build GRCh38
1622685 c.1100C>G p.P367R Chr.7: 20647972-20647972 on Build GRCh38
3024969 c.1118C>T p.S373F Chr.7: 20647990-20647990 on Build GRCh38
4522364 c.1159G>A p.E387K Chr.7: 20648031-20648031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749091082 Chr.7: 20647993-20647993 on Build GRCh38
rs764585719 Chr.7: 20648052-20648052 on Build GRCh38
rs766485384 Chr.7: 20648026-20648026 on Build GRCh38
rs778815031 Chr.7: 20648113-20648113 on Build GRCh38
rs780751423 Chr.7: 20648062-20648062 on Build GRCh38
rs147357794 Chr.7: 20648097-20648097 on Build GRCh38
rs761690956 Chr.7: 20648031-20648031 on Build GRCh38
rs369970820 Chr.7: 20648098-20648098 on Build GRCh38
rs755587897 Chr.7: 20647935-20647935 on Build GRCh38
rs756985564 Chr.7: 20648086-20648086 on Build GRCh38
rs753198026 Chr.7: 20648044-20648044 on Build GRCh38
rs1022750941 Chr.7: 20648075-20648075 on Build GRCh38
rs58976125 Chr.7: 20647977-20647977 on Build GRCh38
rs762905583 Chr.7: 20647945-20647945 on Build GRCh38
rs914576731 Chr.7: 20648109-20648109 on Build GRCh38
rs558475239 Chr.7: 20647982-20647982 on Build GRCh38
rs61732039 Chr.7: 20647981-20647981 on Build GRCh38
rs777303677 Chr.7: 20647973-20647973 on Build GRCh38
rs758721304 Chr.7: 20647964-20647964 on Build GRCh38
rs548174323 Chr.7: 20647958-20647958 on Build GRCh38
rs763353951 Chr.7: 20648051-20648051 on Build GRCh38
rs759646294 Chr.7: 20648032-20648032 on Build GRCh38
rs780722175 Chr.7: 20647979-20647979 on Build GRCh38
rs747845714 Chr.7: 20648118-20648118 on Build GRCh38
rs776923776 Chr.7: 20648124-20648124 on Build GRCh38
rs370636074 Chr.7: 20648122-20648122 on Build GRCh38
rs765843788 Chr.7: 20648037-20648037 on Build GRCh38
rs774007500 Chr.7: 20647999-20647999 on Build GRCh38
rs534432580 Chr.7: 20648017-20648017 on Build GRCh38
rs553136401 Chr.7: 20648095-20648095 on Build GRCh38
rs970197859 Chr.7: 20648082-20648082 on Build GRCh38
rs751840843 Chr.7: 20648083-20648083 on Build GRCh38
rs756853994 Chr.7: 20647978-20647978 on Build GRCh38
rs775369675 Chr.7: 20647985-20647985 on Build GRCh38
rs771212756 Chr.7: 20648120-20648120 on Build GRCh38
rs142733002 Chr.7: 20647994-20647994 on Build GRCh38

View Full Product Details