Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 113240392-113240665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK2
Entrez Gene ID
287
Gene Name
ankyrin 2, neuronal
Gene Aliases
ANK-2, LQT4, brank-2
UniGene
Hs.620557
Gene Chromosome Location
Chr.4: 112706083-113383740 on Build GRCh38
Transcripts
NM_001127493.1 NM_001148.4 NM_020977.3 XM_005262945.2 XM_017008067.1 XM_017008068.1 XM_017008069.1 XM_017008070.1 XM_017008071.1 XM_017008072.1 XM_017008073.1 XM_017008074.1 XM_017008075.1 XM_017008076.1 XM_017008077.1 XM_017008078.1 XM_017008079.1 XM_017008080.1 XM_017008081.1 XM_017008082.1 XM_017008083.1 XM_017008084.1 XM_017008085.1 XM_017008086.1 XM_017008087.1 XM_017008088.1 XM_017008089.1 XM_017008090.1 XM_017008091.1 XM_017008092.1 XM_017008093.1 XM_017008094.1 XM_017008095.1 XM_017008096.1 XM_017008097.1 XM_017008098.1 XM_017008099.1 XM_017008100.1 XM_017008101.1 XM_017008102.1 XM_017008103.1 XM_017008104.1 XM_017008105.1 XM_017008106.1 XM_017008107.1 XM_017008108.1 XM_017008109.1 XM_017008110.1 XM_017008111.1 XM_017008112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1741799 c.754C>T p.L252F Chr.4: 113240545-113240545 on Build GRCh38
3599228 c.709T>C p.L237L Chr.4: 113240500-113240500 on Build GRCh38
4121433 c.649C>A p.H217N Chr.4: 113240503-113240503 on Build GRCh38
6166266 c.682G>C p.A228P Chr.4: 113240536-113240536 on Build GRCh38
4566023 c.729_729+1GG>AA p.? Chr.4: 113240583-113240584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72675204 Chr.4: 113240451-113240451 on Build GRCh38
rs992507931 Chr.4: 113240446-113240446 on Build GRCh38
rs762444022 Chr.4: 113240471-113240471 on Build GRCh38
rs538470141 Chr.4: 113240638-113240638 on Build GRCh38
rs780276854 Chr.4: 113240462-113240462 on Build GRCh38
rs944870683 Chr.4: 113240566-113240566 on Build GRCh38
rs779932254 Chr.4: 113240623-113240623 on Build GRCh38
rs773812842 Chr.4: 113240520-113240520 on Build GRCh38
rs201208094 Chr.4: 113240535-113240535 on Build GRCh38
rs374253227 Chr.4: 113240587-113240587 on Build GRCh38
rs764008198 Chr.4: 113240555-113240555 on Build GRCh38
rs190102899 Chr.4: 113240532-113240532 on Build GRCh38
rs770405996 Chr.4: 113240494-113240494 on Build GRCh38
rs555494739 Chr.4: 113240424-113240424 on Build GRCh38
rs776859965 Chr.4: 113240465-113240465 on Build GRCh38
rs796238253 Chr.4: 113240418-113240418 on Build GRCh38
rs371578231 Chr.4: 113240613-113240613 on Build GRCh38
rs773215795 Chr.4: 113240575-113240575 on Build GRCh38
rs900693189 Chr.4: 113240589-113240589 on Build GRCh38
rs367592681 Chr.4: 113240599-113240599 on Build GRCh38
rs747326413 Chr.4: 113240627-113240627 on Build GRCh38
rs113388306 Chr.4: 113240452-113240452 on Build GRCh38
rs759136608 Chr.4: 113240521-113240521 on Build GRCh38
rs769073818 Chr.4: 113240464-113240464 on Build GRCh38
rs112086259 Chr.4: 113240591-113240591 on Build GRCh38
rs752395758 Chr.4: 113240533-113240533 on Build GRCh38
rs375914552 Chr.4: 113240598-113240598 on Build GRCh38
rs745918500 Chr.4: 113240439-113240439 on Build GRCh38
rs45616931 Chr.4: 113240463-113240463 on Build GRCh38
rs878854260 Chr.4: 113240593-113240593 on Build GRCh38
rs750313251 Chr.4: 113240603-113240603 on Build GRCh38

View Full Product Details