Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122091490-122091759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD86
Entrez Gene ID
942
Gene Name
CD86 molecule
Gene Aliases
B7-2, B7.2, B70, CD28LG2, LAB72
UniGene
Hs.171182
Gene Chromosome Location
Chr.3: 122055362-122121143 on Build GRCh38
Transcripts
NM_001206924.1 NM_001206925.1 NM_006889.4 NM_175862.4 NM_176892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623924 c.50C>T p.A17V Chr.3: 122091636-122091636 on Build GRCh38
5769767 c.53T>C p.F18S Chr.3: 122091639-122091639 on Build GRCh38
3586189 c.22G>A p.G8R Chr.3: 122091608-122091608 on Build GRCh38
7360317 c.50C>G p.A17G Chr.3: 122091636-122091636 on Build GRCh38
3426959 c.60C>A p.L20L Chr.3: 122091646-122091646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751225257 Chr.3: 122091691-122091691 on Build GRCh38
rs904960335 Chr.3: 122091544-122091544 on Build GRCh38
rs377045850 Chr.3: 122091620-122091620 on Build GRCh38
rs113910837 Chr.3: 122091588-122091588 on Build GRCh38
rs760013262 Chr.3: 122091659-122091659 on Build GRCh38
rs756928548 Chr.3: 122091699-122091699 on Build GRCh38
rs760939521 Chr.3: 122091632-122091632 on Build GRCh38
rs915120854 Chr.3: 122091520-122091520 on Build GRCh38
rs770985384 Chr.3: 122091649-122091649 on Build GRCh38
rs869285621 Chr.3: 122091588-122091589 on Build GRCh38
rs763411836 Chr.3: 122091681-122091681 on Build GRCh38
rs768373907 Chr.3: 122091623-122091623 on Build GRCh38
rs955553670 Chr.3: 122091720-122091720 on Build GRCh38
rs1003729667 Chr.3: 122091546-122091546 on Build GRCh38
rs753218784 Chr.3: 122091673-122091673 on Build GRCh38
rs754362665 Chr.3: 122091570-122091570 on Build GRCh38
rs761287421 Chr.3: 122091579-122091579 on Build GRCh38
rs937610131 Chr.3: 122091715-122091715 on Build GRCh38
rs543589700 Chr.3: 122091593-122091593 on Build GRCh38
rs776172845 Chr.3: 122091574-122091574 on Build GRCh38
rs765739697 Chr.3: 122091582-122091582 on Build GRCh38
rs777596388 Chr.3: 122091586-122091586 on Build GRCh38
rs57304749 Chr.3: 122091589-122091589 on Build GRCh38
rs765871942 Chr.3: 122091670-122091670 on Build GRCh38
rs756729117 Chr.3: 122091589-122091589 on Build GRCh38
rs764491301 Chr.3: 122091685-122091685 on Build GRCh38
rs987324645 Chr.3: 122091723-122091723 on Build GRCh38
rs374044667 Chr.3: 122091554-122091554 on Build GRCh38
rs1056206106 Chr.3: 122091547-122091547 on Build GRCh38
rs569458278 Chr.3: 122091521-122091521 on Build GRCh38
rs758149181 Chr.3: 122091670-122091671 on Build GRCh38
rs746602029 Chr.3: 122091587-122091587 on Build GRCh38
rs775312353 Chr.3: 122091611-122091611 on Build GRCh38
rs201456834 Chr.3: 122091658-122091658 on Build GRCh38
rs577864930 Chr.3: 122091572-122091572 on Build GRCh38
rs63114861 Chr.3: 122091581-122091581 on Build GRCh38
rs769737392 Chr.3: 122091595-122091595 on Build GRCh38
rs773436343 Chr.3: 122091625-122091625 on Build GRCh38
rs745767119 Chr.3: 122091615-122091615 on Build GRCh38
rs1052341373 Chr.3: 122091517-122091517 on Build GRCh38
rs753093906 Chr.3: 122091585-122091585 on Build GRCh38
rs557937753 Chr.3: 122091527-122091527 on Build GRCh38
rs765196947 Chr.3: 122091580-122091581 on Build GRCh38
rs1045273266 Chr.3: 122091532-122091532 on Build GRCh38
rs200671146 Chr.3: 122091581-122091581 on Build GRCh38
rs780736216 Chr.3: 122091592-122091592 on Build GRCh38
rs574218976 Chr.3: 122091719-122091719 on Build GRCh38

View Full Product Details