Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 63632935-63633176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL3
Entrez Gene ID
25923
Gene Name
atlastin GTPase 3
Gene Aliases
HSN1F
UniGene
Hs.356719
Gene Chromosome Location
Chr.11: 63624082-63671974 on Build GRCh38
Transcripts
NM_001290048.1 NM_015459.4 XM_006718493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4199188 c.1188+10A>T p.? Chr.11: 63633016-63633016 on Build GRCh38
5839110 c.1069G>A p.A357T Chr.11: 63633064-63633064 on Build GRCh38
5839111 c.1150G>A p.A384T Chr.11: 63633064-63633064 on Build GRCh38
5548109 c.1117-8_1117-7delCT p.? Chr.11: 63633104-63633105 on Build GRCh38
7178542 c.1045G>A p.E349K Chr.11: 63633088-63633088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17656941 Chr.11: 63633021-63633021 on Build GRCh38
rs750304802 Chr.11: 63633131-63633131 on Build GRCh38
rs575525185 Chr.11: 63633076-63633076 on Build GRCh38
rs772075062 Chr.11: 63633050-63633050 on Build GRCh38
rs753601146 Chr.11: 63633141-63633141 on Build GRCh38
rs1035221774 Chr.11: 63633092-63633092 on Build GRCh38
rs911908133 Chr.11: 63632980-63632980 on Build GRCh38
rs563907155 Chr.11: 63633145-63633145 on Build GRCh38
rs764595546 Chr.11: 63633135-63633139 on Build GRCh38
rs1011933757 Chr.11: 63633116-63633116 on Build GRCh38
rs1050984650 Chr.11: 63632973-63632973 on Build GRCh38
rs369978511 Chr.11: 63633022-63633022 on Build GRCh38
rs374642752 Chr.11: 63632966-63632966 on Build GRCh38
rs749140482 Chr.11: 63633016-63633016 on Build GRCh38
rs763776359 Chr.11: 63633103-63633103 on Build GRCh38
rs763532852 Chr.11: 63633090-63633091 on Build GRCh38
rs756770197 Chr.11: 63632995-63632995 on Build GRCh38
rs766943886 Chr.11: 63633127-63633127 on Build GRCh38
rs750394859 Chr.11: 63633008-63633008 on Build GRCh38
rs150194013 Chr.11: 63632993-63632993 on Build GRCh38
rs1037410729 Chr.11: 63632972-63632972 on Build GRCh38
rs747529831 Chr.11: 63633067-63633067 on Build GRCh38
rs902688720 Chr.11: 63633077-63633077 on Build GRCh38
rs761351000 Chr.11: 63633122-63633122 on Build GRCh38
rs777532479 Chr.11: 63633139-63633139 on Build GRCh38
rs762678291 Chr.11: 63632978-63632978 on Build GRCh38
rs773788187 Chr.11: 63633056-63633056 on Build GRCh38
rs758013424 Chr.11: 63633107-63633107 on Build GRCh38
rs781138719 Chr.11: 63633004-63633004 on Build GRCh38
rs200826488 Chr.11: 63633089-63633089 on Build GRCh38
rs139006668 Chr.11: 63633049-63633049 on Build GRCh38
rs777128321 Chr.11: 63633081-63633081 on Build GRCh38
rs966965254 Chr.11: 63633103-63633104 on Build GRCh38
rs756035047 Chr.11: 63633134-63633134 on Build GRCh38
rs773901584 Chr.11: 63633115-63633115 on Build GRCh38
rs201750318 Chr.11: 63633011-63633011 on Build GRCh38

View Full Product Details