Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113274170-113274441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOC
Entrez Gene ID
91653
Gene Name
BOC cell adhesion associated, oncogene regulated
Gene Aliases
CDON2
UniGene
Hs.591318
Gene Chromosome Location
Chr.3: 113211003-113287463 on Build GRCh38
Transcripts
NM_001301861.1 NM_033254.3 XM_005247891.2 XM_005247892.2 XM_005247897.2 XM_011513305.2 XM_017007441.1 XM_017007442.1 XM_017007443.1 XM_017007444.1 XM_017007445.1 XM_017007446.1 XM_017007447.1 XM_017007448.1 XM_017007449.1 XM_017007450.1 XM_017007451.1 XM_017007452.1 XM_017007453.1 XM_017007454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6095731 c.1296C>A p.P432P Chr.3: 113274436-113274436 on Build GRCh38
1752804 c.1292C>T p.S431L Chr.3: 113274432-113274432 on Build GRCh38
1036430 c.1285C>T p.P429S Chr.3: 113274425-113274425 on Build GRCh38
6162663 c.1243C>A p.P415T Chr.3: 113274383-113274383 on Build GRCh38
4583799 c.1236C>T p.S412S Chr.3: 113274376-113274376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140794918 Chr.3: 113274410-113274410 on Build GRCh38
rs759352827 Chr.3: 113274378-113274378 on Build GRCh38
rs1012576047 Chr.3: 113274203-113274203 on Build GRCh38
rs1005871417 Chr.3: 113274228-113274228 on Build GRCh38
rs955831454 Chr.3: 113274280-113274281 on Build GRCh38
rs775595950 Chr.3: 113274387-113274387 on Build GRCh38
rs777946430 Chr.3: 113274305-113274305 on Build GRCh38
rs558920972 Chr.3: 113274251-113274251 on Build GRCh38
rs997359233 Chr.3: 113274274-113274274 on Build GRCh38
rs763887838 Chr.3: 113274321-113274321 on Build GRCh38
rs866012175 Chr.3: 113274375-113274375 on Build GRCh38
rs765304929 Chr.3: 113274379-113274379 on Build GRCh38
rs537795063 Chr.3: 113274420-113274420 on Build GRCh38
rs570385666 Chr.3: 113274405-113274405 on Build GRCh38
rs896627551 Chr.3: 113274208-113274208 on Build GRCh38
rs956663172 Chr.3: 113274294-113274294 on Build GRCh38
rs770611976 Chr.3: 113274332-113274332 on Build GRCh38
rs756361725 Chr.3: 113274411-113274411 on Build GRCh38
rs887480623 Chr.3: 113274219-113274219 on Build GRCh38
rs1030127742 Chr.3: 113274274-113274274 on Build GRCh38
rs926596121 Chr.3: 113274421-113274421 on Build GRCh38
rs988959726 Chr.3: 113274297-113274297 on Build GRCh38
rs369561468 Chr.3: 113274391-113274391 on Build GRCh38
rs533652303 Chr.3: 113274341-113274341 on Build GRCh38
rs746656420 Chr.3: 113274327-113274327 on Build GRCh38
rs1030684296 Chr.3: 113274268-113274268 on Build GRCh38
rs760818320 Chr.3: 113274198-113274198 on Build GRCh38
rs147556461 Chr.3: 113274376-113274376 on Build GRCh38
rs900434562 Chr.3: 113274252-113274252 on Build GRCh38
rs767715937 Chr.3: 113274392-113274392 on Build GRCh38
rs13093350 Chr.3: 113274216-113274216 on Build GRCh38
rs933277105 Chr.3: 113274381-113274381 on Build GRCh38
rs970354374 Chr.3: 113274244-113274244 on Build GRCh38
rs914716046 Chr.3: 113274302-113274302 on Build GRCh38
rs766612660 Chr.3: 113274414-113274414 on Build GRCh38
rs929048731 Chr.3: 113274209-113274209 on Build GRCh38

View Full Product Details