Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66480537-66480776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEAN1
Entrez Gene ID
146227
Gene Name
brain expressed, associated with NEDD4, 1
Gene Aliases
BEAN, SCA31
UniGene
Hs.740218
Gene Chromosome Location
Chr.16: 66427297-66495288 on Build GRCh38
Transcripts
NM_001136106.4 NM_001178020.2 NM_001197224.3 NM_001197225.3 XM_011522883.1 XM_011522884.1 XM_011522885.2 XM_011522888.1 XM_011522889.1 XM_011522890.1 XM_011522891.1 XM_011522892.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
972161 c.531G>A p.T177T Chr.16: 66480676-66480676 on Build GRCh38
5570292 c.530C>T p.T177M Chr.16: 66480675-66480675 on Build GRCh38
6295832 c.534G>T p.L178L Chr.16: 66480679-66480679 on Build GRCh38
6555783 c.494C>T p.A165V Chr.16: 66480639-66480639 on Build GRCh38
1378769 c.611G>A p.G204D Chr.16: 66480756-66480756 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993568312 Chr.16: 66480754-66480754 on Build GRCh38
rs762543522 Chr.16: 66480655-66480655 on Build GRCh38
rs934466849 Chr.16: 66480649-66480649 on Build GRCh38
rs772848546 Chr.16: 66480590-66480590 on Build GRCh38
rs768135296 Chr.16: 66480702-66480702 on Build GRCh38
rs571294840 Chr.16: 66480745-66480745 on Build GRCh38
rs1047457830 Chr.16: 66480720-66480720 on Build GRCh38
rs1041894909 Chr.16: 66480644-66480644 on Build GRCh38
rs530533484 Chr.16: 66480637-66480637 on Build GRCh38
rs780307718 Chr.16: 66480719-66480719 on Build GRCh38
rs541823463 Chr.16: 66480633-66480633 on Build GRCh38
rs141546513 Chr.16: 66480646-66480646 on Build GRCh38
rs1016250921 Chr.16: 66480733-66480733 on Build GRCh38
rs202095246 Chr.16: 66480709-66480709 on Build GRCh38
rs770839939 Chr.16: 66480613-66480613 on Build GRCh38
rs746865919 Chr.16: 66480611-66480611 on Build GRCh38
rs989073790 Chr.16: 66480622-66480622 on Build GRCh38
rs774348951 Chr.16: 66480607-66480607 on Build GRCh38
rs913669034 Chr.16: 66480625-66480625 on Build GRCh38
rs551176359 Chr.16: 66480687-66480687 on Build GRCh38
rs938983764 Chr.16: 66480647-66480647 on Build GRCh38
rs901484789 Chr.16: 66480703-66480703 on Build GRCh38
rs559175464 Chr.16: 66480582-66480582 on Build GRCh38
rs763848920 Chr.16: 66480665-66480665 on Build GRCh38
rs971508505 Chr.16: 66480634-66480634 on Build GRCh38
rs1004333572 Chr.16: 66480744-66480744 on Build GRCh38
rs943247312 Chr.16: 66480691-66480691 on Build GRCh38
rs372012072 Chr.16: 66480581-66480581 on Build GRCh38
rs1054414013 Chr.16: 66480675-66480675 on Build GRCh38
rs1029806340 Chr.16: 66480579-66480579 on Build GRCh38
rs376026948 Chr.16: 66480589-66480589 on Build GRCh38
rs564012020 Chr.16: 66480704-66480704 on Build GRCh38
rs751848971 Chr.16: 66480676-66480676 on Build GRCh38
rs1056453062 Chr.16: 66480654-66480654 on Build GRCh38
rs561638646 Chr.16: 66480636-66480636 on Build GRCh38
rs759655117 Chr.16: 66480718-66480718 on Build GRCh38
rs550452891 Chr.16: 66480692-66480692 on Build GRCh38

View Full Product Details