Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31459643-31459888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC5
Entrez Gene ID
79798
Gene Name
armadillo repeat containing 5
Gene Aliases
AIMAH2
UniGene
Hs.732945
Gene Chromosome Location
Chr.16: 31458273-31467167 on Build GRCh38
Transcripts
NM_001105247.1 NM_001288767.1 NM_001301820.1 NM_024742.2 XM_006721091.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4473267 c.183C>T p.F61F Chr.16: 31459707-31459707 on Build GRCh38
6429789 c.524C>T p.A175V Chr.16: 31459763-31459763 on Build GRCh38
3509241 c.617C>T p.P206L Chr.16: 31459856-31459856 on Build GRCh38
4489470 c.632C>T p.S211L Chr.16: 31459871-31459871 on Build GRCh38
5850824 c.450delG p.G152fs*80 Chr.16: 31459689-31459689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1041411723 Chr.16: 31459703-31459703 on Build GRCh38
rs976862231 Chr.16: 31459854-31459854 on Build GRCh38
rs998495351 Chr.16: 31459753-31459753 on Build GRCh38
rs1049912614 Chr.16: 31459696-31459696 on Build GRCh38
rs538649451 Chr.16: 31459866-31459868 on Build GRCh38
rs758689001 Chr.16: 31459784-31459784 on Build GRCh38
rs766946870 Chr.16: 31459755-31459755 on Build GRCh38
rs1008632310 Chr.16: 31459701-31459701 on Build GRCh38
rs762616028 Chr.16: 31459663-31459663 on Build GRCh38
rs1050026325 Chr.16: 31459864-31459864 on Build GRCh38
rs750844063 Chr.16: 31459781-31459781 on Build GRCh38
rs184376263 Chr.16: 31459866-31459866 on Build GRCh38
rs778311307 Chr.16: 31459682-31459682 on Build GRCh38
rs1003301079 Chr.16: 31459816-31459816 on Build GRCh38
rs373860441 Chr.16: 31459861-31459861 on Build GRCh38
rs756248578 Chr.16: 31459853-31459853 on Build GRCh38
rs777779636 Chr.16: 31459858-31459858 on Build GRCh38
rs758286564 Chr.16: 31459771-31459771 on Build GRCh38
rs765712803 Chr.16: 31459776-31459776 on Build GRCh38
rs1016610213 Chr.16: 31459829-31459846 on Build GRCh38
rs1033133219 Chr.16: 31459770-31459770 on Build GRCh38
rs951869246 Chr.16: 31459688-31459689 on Build GRCh38
rs983210811 Chr.16: 31459843-31459843 on Build GRCh38
rs562540127 Chr.16: 31459812-31459812 on Build GRCh38
rs550452188 Chr.16: 31459714-31459714 on Build GRCh38
rs181081811 Chr.16: 31459698-31459698 on Build GRCh38
rs781368727 Chr.16: 31459800-31459800 on Build GRCh38
rs766609669 Chr.16: 31459846-31459846 on Build GRCh38
rs750129420 Chr.16: 31459662-31459662 on Build GRCh38
rs748216236 Chr.16: 31459805-31459805 on Build GRCh38
rs759061318 Chr.16: 31459749-31459749 on Build GRCh38
rs745352803 Chr.16: 31459811-31459837 on Build GRCh38
rs771753877 Chr.16: 31459868-31459868 on Build GRCh38
rs770791789 Chr.16: 31459863-31459863 on Build GRCh38
rs1022735732 Chr.16: 31459803-31459803 on Build GRCh38
rs751157724 Chr.16: 31459732-31459732 on Build GRCh38
rs750885418 Chr.16: 31459839-31459839 on Build GRCh38
rs773106431 Chr.16: 31459716-31459716 on Build GRCh38
rs1018679982 Chr.16: 31459729-31459729 on Build GRCh38
rs932682237 Chr.16: 31459779-31459779 on Build GRCh38
rs747183399 Chr.16: 31459694-31459694 on Build GRCh38
rs757715132 Chr.16: 31459767-31459767 on Build GRCh38
rs775123648 Chr.16: 31459869-31459869 on Build GRCh38
rs780112907 Chr.16: 31459691-31459691 on Build GRCh38
rs766121561 Chr.16: 31459722-31459722 on Build GRCh38
rs769890229 Chr.16: 31459806-31459806 on Build GRCh38
rs765844255 Chr.16: 31459836-31459836 on Build GRCh38
rs758790821 Chr.16: 31459842-31459842 on Build GRCh38
rs894075781 Chr.16: 31459791-31459791 on Build GRCh38
rs199693319 Chr.16: 31459867-31459867 on Build GRCh38
rs548290271 Chr.16: 31459850-31459850 on Build GRCh38
rs201768837 Chr.16: 31459852-31459852 on Build GRCh38
rs760240565 Chr.16: 31459870-31459870 on Build GRCh38
rs749164561 Chr.16: 31459811-31459811 on Build GRCh38
rs751120534 Chr.16: 31459671-31459671 on Build GRCh38
rs773018104 Chr.16: 31459808-31459808 on Build GRCh38
rs754286092 Chr.16: 31459688-31459688 on Build GRCh38
rs757625031 Chr.16: 31459689-31459689 on Build GRCh38
rs780333333 Chr.16: 31459788-31459788 on Build GRCh38
rs532321693 Chr.16: 31459700-31459700 on Build GRCh38
rs762622578 Chr.16: 31459718-31459718 on Build GRCh38
rs587777660 Chr.16: 31459780-31459780 on Build GRCh38
rs765924062 Chr.16: 31459667-31459667 on Build GRCh38
rs754376008 Chr.16: 31459765-31459765 on Build GRCh38
rs760150629 Chr.16: 31459832-31459832 on Build GRCh38
rs932874365 Chr.16: 31459857-31459857 on Build GRCh38
rs372292806 Chr.16: 31459818-31459818 on Build GRCh38
rs571045241 Chr.16: 31459871-31459871 on Build GRCh38
rs767038404 Chr.16: 31459821-31459821 on Build GRCh38
rs755137110 Chr.16: 31459794-31459794 on Build GRCh38
rs747130429 Chr.16: 31459793-31459793 on Build GRCh38
rs772390098 Chr.16: 31459690-31459690 on Build GRCh38

View Full Product Details