Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 191380769-191380994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC50
Entrez Gene ID
152137
Gene Name
coiled-coil domain containing 50
Gene Aliases
C3orf6, DFNA44, YMER
UniGene
Hs.478682
Gene Chromosome Location
Chr.3: 191329082-191398670 on Build GRCh38
Transcripts
NM_174908.3 NM_178335.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7359962 c.1181A>G p.K394R Chr.3: 191380871-191380871 on Build GRCh38
1042559 c.1242+2T>C p.? Chr.3: 191380934-191380934 on Build GRCh38
171256 c.1148G>A p.R383Q Chr.3: 191380838-191380838 on Build GRCh38
3333389 c.1200G>T p.E400D Chr.3: 191380890-191380890 on Build GRCh38
7359964 c.1144G>A p.A382T Chr.3: 191380834-191380834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769063488 Chr.3: 191380928-191380928 on Build GRCh38
rs145988140 Chr.3: 191380906-191380906 on Build GRCh38
rs979845402 Chr.3: 191380800-191380800 on Build GRCh38
rs773033895 Chr.3: 191380950-191380950 on Build GRCh38
rs768923251 Chr.3: 191380852-191380852 on Build GRCh38
rs757073067 Chr.3: 191380947-191380948 on Build GRCh38
rs750383173 Chr.3: 191380798-191380798 on Build GRCh38
rs762479219 Chr.3: 191380955-191380955 on Build GRCh38
rs757529140 Chr.3: 191380897-191380897 on Build GRCh38
rs774280604 Chr.3: 191380961-191380961 on Build GRCh38
rs900748864 Chr.3: 191380925-191380925 on Build GRCh38
rs746357460 Chr.3: 191380840-191380840 on Build GRCh38
rs753822249 Chr.3: 191380833-191380833 on Build GRCh38
rs769610982 Chr.3: 191380947-191380947 on Build GRCh38
rs766303798 Chr.3: 191380877-191380877 on Build GRCh38
rs772898236 Chr.3: 191380876-191380876 on Build GRCh38
rs775687066 Chr.3: 191380847-191380847 on Build GRCh38
rs758438006 Chr.3: 191380922-191380922 on Build GRCh38
rs1039059170 Chr.3: 191380894-191380894 on Build GRCh38
rs910796129 Chr.3: 191380886-191380886 on Build GRCh38
rs765824309 Chr.3: 191380959-191380959 on Build GRCh38
rs536917876 Chr.3: 191380924-191380924 on Build GRCh38
rs146511344 Chr.3: 191380893-191380893 on Build GRCh38
rs751414977 Chr.3: 191380812-191380812 on Build GRCh38
rs377042068 Chr.3: 191380885-191380885 on Build GRCh38
rs138707536 Chr.3: 191380921-191380921 on Build GRCh38
rs211043 Chr.3: 191380942-191380942 on Build GRCh38
rs779692812 Chr.3: 191380809-191380809 on Build GRCh38
rs780560171 Chr.3: 191380923-191380923 on Build GRCh38
rs767218061 Chr.3: 191380888-191380888 on Build GRCh38
rs758468377 Chr.3: 191380808-191380808 on Build GRCh38
rs772679997 Chr.3: 191380846-191380846 on Build GRCh38
rs779028705 Chr.3: 191380838-191380838 on Build GRCh38
rs978124534 Chr.3: 191380849-191380849 on Build GRCh38
rs776357824 Chr.3: 191380854-191380854 on Build GRCh38
rs761626192 Chr.3: 191380867-191380867 on Build GRCh38
rs753560713 Chr.3: 191380806-191380807 on Build GRCh38
rs143628723 Chr.3: 191380834-191380834 on Build GRCh38
rs727504888 Chr.3: 191380911-191380911 on Build GRCh38
rs114502673 Chr.3: 191380871-191380871 on Build GRCh38

View Full Product Details