Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48645253-48645516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA11
Entrez Gene ID
770
Gene Name
carbonic anhydrase 11
Gene Aliases
CARPX1
UniGene
Hs.428446
Gene Chromosome Location
Chr.19: 48637942-48646312 on Build GRCh38
Transcripts
NM_001217.4 NR_136241.1 NR_004401.2

Gene Symbol
SEC1P
Entrez Gene ID
653677
Gene Name
secretory blood group 1, pseudogene
Gene Aliases
SEC1
UniGene
Hs.443160
Gene Chromosome Location
Chr.19: 48638039-48682245 on Build GRCh38
Transcripts
NM_001217.4 NR_136241.1 NR_004401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5497436 c.119delA p.N40fs*16 Chr.19: 48645426-48645426 on Build GRCh38
7329510 c.93_94insC p.E32fs*9 Chr.19: 48645451-48645452 on Build GRCh38
7358827 c.82G>A p.A28T Chr.19: 48645463-48645463 on Build GRCh38
4522178 c.114G>A p.K38K Chr.19: 48645431-48645431 on Build GRCh38
6322321 c.99C>A p.D33E Chr.19: 48645446-48645446 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1128668 Chr.19: 48645475-48645475 on Build GRCh38
rs886941353 Chr.19: 48645350-48645352 on Build GRCh38
rs1029380930 Chr.19: 48645455-48645455 on Build GRCh38
rs200820630 Chr.19: 48645470-48645470 on Build GRCh38
rs551658222 Chr.19: 48645312-48645312 on Build GRCh38
rs1012532111 Chr.19: 48645311-48645311 on Build GRCh38
rs570511339 Chr.19: 48645313-48645313 on Build GRCh38
rs570262876 Chr.19: 48645387-48645387 on Build GRCh38
rs1048958523 Chr.19: 48645292-48645292 on Build GRCh38
rs779405514 Chr.19: 48645328-48645328 on Build GRCh38
rs998522989 Chr.19: 48645357-48645357 on Build GRCh38
rs766344020 Chr.19: 48645466-48645466 on Build GRCh38
rs549668589 Chr.19: 48645390-48645390 on Build GRCh38
rs753983346 Chr.19: 48645281-48645281 on Build GRCh38
rs371325088 Chr.19: 48645452-48645452 on Build GRCh38
rs1128667 Chr.19: 48645477-48645477 on Build GRCh38
rs954066288 Chr.19: 48645419-48645419 on Build GRCh38
rs746031117 Chr.19: 48645356-48645356 on Build GRCh38
rs957534518 Chr.19: 48645383-48645383 on Build GRCh38
rs772965243 Chr.19: 48645410-48645410 on Build GRCh38
rs182731741 Chr.19: 48645287-48645287 on Build GRCh38
rs758849958 Chr.19: 48645364-48645364 on Build GRCh38
rs146677575 Chr.19: 48645472-48645472 on Build GRCh38
rs961471106 Chr.19: 48645496-48645496 on Build GRCh38
rs551950382 Chr.19: 48645310-48645310 on Build GRCh38
rs774274941 Chr.19: 48645469-48645469 on Build GRCh38
rs116709645 Chr.19: 48645389-48645389 on Build GRCh38
rs369858296 Chr.19: 48645406-48645406 on Build GRCh38
rs144597404 Chr.19: 48645448-48645448 on Build GRCh38
rs896540792 Chr.19: 48645309-48645310 on Build GRCh38
rs747721976 Chr.19: 48645391-48645391 on Build GRCh38
rs1056579066 Chr.19: 48645285-48645285 on Build GRCh38
rs764143099 Chr.19: 48645274-48645274 on Build GRCh38
rs986791956 Chr.19: 48645449-48645449 on Build GRCh38
rs866139598 Chr.19: 48645425-48645425 on Build GRCh38
rs996894535 Chr.19: 48645414-48645414 on Build GRCh38

View Full Product Details