Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 25136098-25136351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf31
Entrez Gene ID
136895
Gene Name
chromosome 7 open reading frame 31
Gene Aliases
-
UniGene
Hs.122055
Gene Chromosome Location
Chr.7: 25134697-25180748 on Build GRCh38
Transcripts
NM_138811.3 XM_005249619.3 XM_005249620.3 XM_011515125.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5603007 c.1588C>T p.P530S Chr.7: 25136157-25136157 on Build GRCh38
4619875 c.1580G>C p.G527A Chr.7: 25136165-25136165 on Build GRCh38
3785074 c.1441C>A p.P481T Chr.7: 25136304-25136304 on Build GRCh38
5674600 c.1527A>G p.E509E Chr.7: 25136218-25136218 on Build GRCh38
236374 c.1480C>A p.Q494K Chr.7: 25136265-25136265 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775080372 Chr.7: 25136302-25136303 on Build GRCh38
rs766882133 Chr.7: 25136274-25136274 on Build GRCh38
rs987917875 Chr.7: 25136315-25136319 on Build GRCh38
rs768367867 Chr.7: 25136270-25136270 on Build GRCh38
rs751375207 Chr.7: 25136319-25136319 on Build GRCh38
rs17857512 Chr.7: 25136193-25136193 on Build GRCh38
rs760112679 Chr.7: 25136285-25136285 on Build GRCh38
rs200916847 Chr.7: 25136318-25136318 on Build GRCh38
rs746838447 Chr.7: 25136224-25136227 on Build GRCh38
rs767817194 Chr.7: 25136137-25136137 on Build GRCh38
rs768801754 Chr.7: 25136187-25136188 on Build GRCh38
rs147311660 Chr.7: 25136258-25136258 on Build GRCh38
rs1043022419 Chr.7: 25136135-25136135 on Build GRCh38
rs761170590 Chr.7: 25136153-25136153 on Build GRCh38
rs765391946 Chr.7: 25136190-25136190 on Build GRCh38
rs776475768 Chr.7: 25136240-25136241 on Build GRCh38
rs757205010 Chr.7: 25136248-25136248 on Build GRCh38
rs199628566 Chr.7: 25136131-25136131 on Build GRCh38
rs572492752 Chr.7: 25136272-25136272 on Build GRCh38
rs201712394 Chr.7: 25136243-25136243 on Build GRCh38
rs375798433 Chr.7: 25136172-25136172 on Build GRCh38
rs759479399 Chr.7: 25136243-25136243 on Build GRCh38
rs758688102 Chr.7: 25136313-25136313 on Build GRCh38
rs775042926 Chr.7: 25136126-25136126 on Build GRCh38
rs765172989 Chr.7: 25136286-25136286 on Build GRCh38
rs139517301 Chr.7: 25136238-25136238 on Build GRCh38
rs764861196 Chr.7: 25136270-25136270 on Build GRCh38
rs774555525 Chr.7: 25136212-25136214 on Build GRCh38
rs143515344 Chr.7: 25136257-25136257 on Build GRCh38
rs745351568 Chr.7: 25136253-25136253 on Build GRCh38
rs76016318 Chr.7: 25136169-25136169 on Build GRCh38
rs190007414 Chr.7: 25136251-25136251 on Build GRCh38
rs895576006 Chr.7: 25136189-25136189 on Build GRCh38
rs912214490 Chr.7: 25136320-25136320 on Build GRCh38
rs956435652 Chr.7: 25136315-25136316 on Build GRCh38
rs34005449 Chr.7: 25136266-25136266 on Build GRCh38
rs528617265 Chr.7: 25136223-25136223 on Build GRCh38
rs1004955475 Chr.7: 25136168-25136168 on Build GRCh38
rs752830224 Chr.7: 25136290-25136290 on Build GRCh38
rs773575593 Chr.7: 25136147-25136147 on Build GRCh38
rs773557047 Chr.7: 25136271-25136271 on Build GRCh38
rs764468781 Chr.7: 25136316-25136316 on Build GRCh38
rs200403535 Chr.7: 25136170-25136170 on Build GRCh38
rs553406677 Chr.7: 25136230-25136231 on Build GRCh38
rs568707268 Chr.7: 25136151-25136151 on Build GRCh38

View Full Product Details