Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66341210-66341441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRMS1
Entrez Gene ID
25855
Gene Name
breast cancer metastasis suppressor 1
Gene Aliases
-
UniGene
Hs.100426
Gene Chromosome Location
Chr.11: 66337333-66345111 on Build GRCh38
Transcripts
NM_001024957.1 NM_015399.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1298452 c.306G>A p.T102T Chr.11: 66341258-66341258 on Build GRCh38
4035687 c.294C>T p.A98A Chr.11: 66341270-66341270 on Build GRCh38
6612891 c.256C>T p.L86L Chr.11: 66341308-66341308 on Build GRCh38
7100052 c.266G>A p.R89Q Chr.11: 66341298-66341298 on Build GRCh38
6612890 c.256C>T p.L86L Chr.11: 66341308-66341308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762502004 Chr.11: 66341259-66341259 on Build GRCh38
rs148062689 Chr.11: 66341244-66341244 on Build GRCh38
rs750862445 Chr.11: 66341285-66341285 on Build GRCh38
rs375945773 Chr.11: 66341378-66341378 on Build GRCh38
rs371890921 Chr.11: 66341231-66341231 on Build GRCh38
rs772670092 Chr.11: 66341370-66341370 on Build GRCh38
rs746612499 Chr.11: 66341349-66341349 on Build GRCh38
rs980544403 Chr.11: 66341270-66341270 on Build GRCh38
rs745705747 Chr.11: 66341313-66341313 on Build GRCh38
rs199933125 Chr.11: 66341292-66341292 on Build GRCh38
rs564172836 Chr.11: 66341298-66341298 on Build GRCh38
rs746996980 Chr.11: 66341235-66341235 on Build GRCh38
rs765437527 Chr.11: 66341242-66341243 on Build GRCh38
rs760946811 Chr.11: 66341286-66341286 on Build GRCh38
rs757278464 Chr.11: 66341305-66341305 on Build GRCh38
rs762218512 Chr.11: 66341355-66341355 on Build GRCh38
rs367854116 Chr.11: 66341299-66341299 on Build GRCh38
rs374007540 Chr.11: 66341247-66341247 on Build GRCh38
rs774685217 Chr.11: 66341271-66341271 on Build GRCh38
rs763740010 Chr.11: 66341400-66341400 on Build GRCh38
rs932879158 Chr.11: 66341414-66341414 on Build GRCh38
rs773928682 Chr.11: 66341338-66341338 on Build GRCh38
rs943682151 Chr.11: 66341342-66341342 on Build GRCh38
rs768179699 Chr.11: 66341261-66341261 on Build GRCh38
rs976272867 Chr.11: 66341385-66341385 on Build GRCh38
rs894284057 Chr.11: 66341241-66341241 on Build GRCh38
rs777309913 Chr.11: 66341320-66341320 on Build GRCh38
rs748973710 Chr.11: 66341337-66341337 on Build GRCh38
rs567277375 Chr.11: 66341422-66341422 on Build GRCh38
rs1004875942 Chr.11: 66341356-66341356 on Build GRCh38
rs752980531 Chr.11: 66341356-66341357 on Build GRCh38
rs762390912 Chr.11: 66341279-66341279 on Build GRCh38
rs867762827 Chr.11: 66341286-66341292 on Build GRCh38
rs745495658 Chr.11: 66341245-66341245 on Build GRCh38
rs1027722053 Chr.11: 66341248-66341248 on Build GRCh38
rs774951857 Chr.11: 66341250-66341250 on Build GRCh38
rs780845767 Chr.11: 66341308-66341308 on Build GRCh38
rs770564610 Chr.11: 66341243-66341243 on Build GRCh38
rs371465036 Chr.11: 66341372-66341372 on Build GRCh38
rs761069954 Chr.11: 66341377-66341377 on Build GRCh38
rs769414940 Chr.11: 66341315-66341315 on Build GRCh38
rs767755816 Chr.11: 66341281-66341281 on Build GRCh38
rs779635935 Chr.11: 66341319-66341319 on Build GRCh38
rs2236679 Chr.11: 66341423-66341423 on Build GRCh38
rs754134260 Chr.11: 66341294-66341294 on Build GRCh38
rs547533348 Chr.11: 66341420-66341420 on Build GRCh38
rs751394775 Chr.11: 66341304-66341304 on Build GRCh38

View Full Product Details