Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94885476-94885719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNE2
Entrez Gene ID
9134
Gene Name
cyclin E2
Gene Aliases
CYCE2
UniGene
Hs.521693
Gene Chromosome Location
Chr.8: 94880224-94896671 on Build GRCh38
Transcripts
NM_057749.2 XM_011517366.2 XM_017013957.1 XM_017013958.1 XM_017013959.1 XM_017013616.1

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
UniGene
Hs.727669
Gene Chromosome Location
Chr.8: 94823290-94894664 on Build GRCh38
Transcripts
NM_057749.2 XM_011517366.2 XM_017013957.1 XM_017013958.1 XM_017013959.1 XM_017013616.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3652021 c.613C>T p.P205S Chr.8: 94885546-94885546 on Build GRCh38
7089825 c.601-1G>T p.? Chr.8: 94885559-94885559 on Build GRCh38
5730860 c.636C>T p.Y212Y Chr.8: 94885523-94885523 on Build GRCh38
5026950 c.665A>G p.D222G Chr.8: 94885494-94885494 on Build GRCh38
5771375 c.619C>T p.L207F Chr.8: 94885540-94885540 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs574062611 Chr.8: 94885634-94885634 on Build GRCh38
rs113263061 Chr.8: 94885570-94885570 on Build GRCh38
rs140734465 Chr.8: 94885599-94885599 on Build GRCh38
rs771395255 Chr.8: 94885537-94885537 on Build GRCh38
rs761769678 Chr.8: 94885580-94885580 on Build GRCh38
rs750229108 Chr.8: 94885598-94885598 on Build GRCh38
rs764962884 Chr.8: 94885588-94885588 on Build GRCh38
rs535199145 Chr.8: 94885522-94885522 on Build GRCh38
rs184241751 Chr.8: 94885566-94885566 on Build GRCh38
rs902867658 Chr.8: 94885595-94885595 on Build GRCh38
rs760228121 Chr.8: 94885658-94885658 on Build GRCh38
rs555383126 Chr.8: 94885529-94885529 on Build GRCh38
rs757918416 Chr.8: 94885610-94885610 on Build GRCh38
rs564061904 Chr.8: 94885623-94885623 on Build GRCh38
rs887823610 Chr.8: 94885517-94885517 on Build GRCh38
rs202056359 Chr.8: 94885543-94885543 on Build GRCh38
rs764126618 Chr.8: 94885568-94885568 on Build GRCh38
rs368534515 Chr.8: 94885573-94885573 on Build GRCh38
rs957341312 Chr.8: 94885633-94885633 on Build GRCh38
rs759784230 Chr.8: 94885544-94885544 on Build GRCh38
rs574878499 Chr.8: 94885548-94885548 on Build GRCh38
rs998888797 Chr.8: 94885620-94885620 on Build GRCh38
rs775366582 Chr.8: 94885561-94885561 on Build GRCh38

View Full Product Details