Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120660676-120660925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP1
Entrez Gene ID
9478
Gene Name
calcium binding protein 1
Gene Aliases
CALBRAIN, HCALB_BR
UniGene
Hs.458482
Gene Chromosome Location
Chr.12: 120640552-120680869 on Build GRCh38
Transcripts
NM_001033677.1 NM_004276.4 NM_031205.3 XM_017020235.1 XM_017020236.1 XM_017020237.1 XM_017020238.1 XM_017020239.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6015200 c.401-10C>T p.? Chr.12: 120660721-120660721 on Build GRCh38
935953 c.429C>T p.F143F Chr.12: 120660759-120660759 on Build GRCh38
3457061 c.882C>T p.L294L Chr.12: 120660783-120660783 on Build GRCh38
935952 c.858C>T p.F286F Chr.12: 120660759-120660759 on Build GRCh38
3457059 c.844G>A p.D282N Chr.12: 120660745-120660745 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1013011313 Chr.12: 120660883-120660883 on Build GRCh38
rs749605292 Chr.12: 120660864-120660864 on Build GRCh38
rs779594033 Chr.12: 120660848-120660848 on Build GRCh38
rs767718911 Chr.12: 120660710-120660711 on Build GRCh38
rs759463784 Chr.12: 120660760-120660760 on Build GRCh38
rs745312563 Chr.12: 120660698-120660698 on Build GRCh38
rs770332473 Chr.12: 120660733-120660733 on Build GRCh38
rs756019140 Chr.12: 120660797-120660797 on Build GRCh38
rs1022596307 Chr.12: 120660900-120660900 on Build GRCh38
rs762260379 Chr.12: 120660719-120660719 on Build GRCh38
rs752524118 Chr.12: 120660783-120660783 on Build GRCh38
rs1037749606 Chr.12: 120660820-120660820 on Build GRCh38
rs202165204 Chr.12: 120660732-120660732 on Build GRCh38
rs748706808 Chr.12: 120660849-120660849 on Build GRCh38
rs767519753 Chr.12: 120660763-120660763 on Build GRCh38
rs753066924 Chr.12: 120660714-120660715 on Build GRCh38
rs1054705171 Chr.12: 120660888-120660888 on Build GRCh38
rs770420225 Chr.12: 120660856-120660856 on Build GRCh38
rs777033084 Chr.12: 120660846-120660846 on Build GRCh38
rs773679995 Chr.12: 120660740-120660740 on Build GRCh38
rs763333780 Chr.12: 120660741-120660741 on Build GRCh38
rs751671065 Chr.12: 120660753-120660753 on Build GRCh38
rs1013461959 Chr.12: 120660893-120660893 on Build GRCh38
rs753621687 Chr.12: 120660825-120660825 on Build GRCh38
rs893347044 Chr.12: 120660892-120660892 on Build GRCh38
rs898034653 Chr.12: 120660829-120660829 on Build GRCh38
rs762119586 Chr.12: 120660862-120660862 on Build GRCh38
rs374336059 Chr.12: 120660870-120660870 on Build GRCh38
rs778366806 Chr.12: 120660844-120660844 on Build GRCh38
rs769211148 Chr.12: 120660699-120660699 on Build GRCh38
rs368236303 Chr.12: 120660875-120660875 on Build GRCh38
rs774393866 Chr.12: 120660871-120660871 on Build GRCh38
rs777615351 Chr.12: 120660824-120660824 on Build GRCh38
rs372619642 Chr.12: 120660747-120660747 on Build GRCh38
rs145655068 Chr.12: 120660836-120660836 on Build GRCh38
rs745417016 Chr.12: 120660845-120660845 on Build GRCh38
rs776966137 Chr.12: 120660703-120660703 on Build GRCh38
rs552129817 Chr.12: 120660894-120660894 on Build GRCh38
rs767540812 Chr.12: 120660876-120660876 on Build GRCh38

View Full Product Details