Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130207718-130207951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSMEM1
Entrez Gene ID
136263
Gene Name
serine rich single-pass membrane protein 1
Gene Aliases
C7orf45
UniGene
Hs.351816
Gene Chromosome Location
Chr.7: 130205399-130216869 on Build GRCh38
Transcripts
NM_145268.3 XM_005250141.3 XM_011515795.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4934383 c.25T>G p.W9G Chr.7: 130207935-130207935 on Build GRCh38
3633242 c.28G>A p.E10K Chr.7: 130207938-130207938 on Build GRCh38
5583340 c.37C>T p.P13S Chr.7: 130207947-130207947 on Build GRCh38
6176159 c.11T>C p.L4P Chr.7: 130207921-130207921 on Build GRCh38
1448041 c.30delG p.V11fs*1 Chr.7: 130207940-130207940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530894734 Chr.7: 130207838-130207838 on Build GRCh38
rs141272715 Chr.7: 130207826-130207826 on Build GRCh38
rs749707311 Chr.7: 130207920-130207920 on Build GRCh38
rs750500555 Chr.7: 130207917-130207917 on Build GRCh38
rs762624408 Chr.7: 130207899-130207899 on Build GRCh38
rs775993152 Chr.7: 130207767-130207767 on Build GRCh38
rs779288452 Chr.7: 130207924-130207924 on Build GRCh38
rs752584235 Chr.7: 130207892-130207892 on Build GRCh38
rs994684456 Chr.7: 130207896-130207896 on Build GRCh38
rs770216670 Chr.7: 130207755-130207755 on Build GRCh38
rs776224014 Chr.7: 130207863-130207863 on Build GRCh38
rs746384178 Chr.7: 130207749-130207749 on Build GRCh38
rs771809854 Chr.7: 130207782-130207782 on Build GRCh38
rs569062330 Chr.7: 130207834-130207834 on Build GRCh38
rs999435351 Chr.7: 130207758-130207758 on Build GRCh38
rs770588079 Chr.7: 130207811-130207811 on Build GRCh38
rs772486537 Chr.7: 130207777-130207777 on Build GRCh38
rs756675106 Chr.7: 130207919-130207919 on Build GRCh38
rs781251921 Chr.7: 130207911-130207911 on Build GRCh38
rs748641003 Chr.7: 130207750-130207750 on Build GRCh38
rs897723085 Chr.7: 130207886-130207886 on Build GRCh38
rs146270376 Chr.7: 130207903-130207903 on Build GRCh38
rs759552553 Chr.7: 130207864-130207864 on Build GRCh38
rs1007677012 Chr.7: 130207832-130207832 on Build GRCh38
rs760364348 Chr.7: 130207804-130207804 on Build GRCh38
rs932692157 Chr.7: 130207806-130207806 on Build GRCh38
rs977299185 Chr.7: 130207788-130207788 on Build GRCh38
rs765251448 Chr.7: 130207885-130207885 on Build GRCh38
rs772713611 Chr.7: 130207787-130207787 on Build GRCh38
rs757409752 Chr.7: 130207906-130207906 on Build GRCh38
rs572884266 Chr.7: 130207772-130207772 on Build GRCh38
rs921452927 Chr.7: 130207798-130207798 on Build GRCh38

View Full Product Details