Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53306909-53307182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf10
Entrez Gene ID
60314
Gene Name
chromosome 12 open reading frame 10
Gene Aliases
Gamm1, MST024, MSTP024, MYG, MYG1
UniGene
Hs.655988
Gene Chromosome Location
Chr.12: 53299686-53307181 on Build GRCh38
Transcripts
NM_021640.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3998855 c.1050C>T p.G350G Chr.12: 53307068-53307068 on Build GRCh38
468583 c.1019_1020GC>AA p.C340>? Chr.12: 53307037-53307038 on Build GRCh38
468582 c.1019G>A p.C340Y Chr.12: 53307037-53307037 on Build GRCh38
3721008 c.1058A>G p.H353R Chr.12: 53307076-53307076 on Build GRCh38
4666692 c.1057C>T p.H353Y Chr.12: 53307075-53307075 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760920264 Chr.12: 53307063-53307063 on Build GRCh38
rs780741684 Chr.12: 53307084-53307084 on Build GRCh38
rs749637560 Chr.12: 53307088-53307088 on Build GRCh38
rs770202277 Chr.12: 53306955-53306955 on Build GRCh38
rs746731363 Chr.12: 53307026-53307026 on Build GRCh38
rs1011116825 Chr.12: 53307154-53307154 on Build GRCh38
rs367601830 Chr.12: 53307017-53307017 on Build GRCh38
rs752688674 Chr.12: 53307075-53307075 on Build GRCh38
rs142952424 Chr.12: 53306998-53306998 on Build GRCh38
rs553968048 Chr.12: 53307056-53307056 on Build GRCh38
rs754195181 Chr.12: 53307139-53307139 on Build GRCh38
rs771151643 Chr.12: 53307135-53307135 on Build GRCh38
rs773373546 Chr.12: 53307124-53307124 on Build GRCh38
rs775866911 Chr.12: 53307030-53307030 on Build GRCh38
rs764525199 Chr.12: 53306972-53306972 on Build GRCh38
rs747626617 Chr.12: 53307003-53307003 on Build GRCh38
rs749640109 Chr.12: 53307045-53307045 on Build GRCh38
rs763451616 Chr.12: 53306971-53306971 on Build GRCh38
rs374635731 Chr.12: 53306991-53306991 on Build GRCh38
rs368122295 Chr.12: 53307157-53307157 on Build GRCh38
rs151092802 Chr.12: 53307049-53307049 on Build GRCh38
rs138116397 Chr.12: 53307106-53307106 on Build GRCh38
rs762020423 Chr.12: 53306984-53306984 on Build GRCh38
rs150400421 Chr.12: 53307079-53307079 on Build GRCh38
rs756887812 Chr.12: 53307082-53307082 on Build GRCh38
rs711316 Chr.12: 53307076-53307076 on Build GRCh38
rs766129380 Chr.12: 53306992-53306992 on Build GRCh38
rs149538988 Chr.12: 53307114-53307114 on Build GRCh38
rs750530813 Chr.12: 53306935-53306935 on Build GRCh38
rs965236600 Chr.12: 53307091-53307091 on Build GRCh38
rs202213306 Chr.12: 53306988-53306988 on Build GRCh38
rs371647277 Chr.12: 53307068-53307068 on Build GRCh38
rs746208275 Chr.12: 53306946-53306946 on Build GRCh38
rs772891157 Chr.12: 53306939-53306939 on Build GRCh38
rs765198186 Chr.12: 53307070-53307070 on Build GRCh38
rs879399161 Chr.12: 53307128-53307129 on Build GRCh38
rs776767331 Chr.12: 53307145-53307145 on Build GRCh38
rs778177423 Chr.12: 53307051-53307052 on Build GRCh38
rs538724342 Chr.12: 53307047-53307047 on Build GRCh38
rs768954010 Chr.12: 53307095-53307095 on Build GRCh38
rs376729554 Chr.12: 53307081-53307081 on Build GRCh38
rs774443884 Chr.12: 53306981-53306981 on Build GRCh38
rs1045141 Chr.12: 53307074-53307074 on Build GRCh38
rs772716522 Chr.12: 53307123-53307123 on Build GRCh38
rs1003292900 Chr.12: 53307122-53307122 on Build GRCh38
rs142953883 Chr.12: 53307069-53307069 on Build GRCh38
rs760699090 Chr.12: 53307131-53307131 on Build GRCh38
rs758886940 Chr.12: 53307152-53307152 on Build GRCh38
rs752494374 Chr.12: 53307160-53307160 on Build GRCh38
rs775858998 Chr.12: 53306965-53306965 on Build GRCh38
rs757951996 Chr.12: 53307007-53307007 on Build GRCh38
rs759360910 Chr.12: 53307148-53307148 on Build GRCh38
rs372883926 Chr.12: 53306989-53306989 on Build GRCh38
rs770496861 Chr.12: 53307029-53307029 on Build GRCh38
rs142504669 Chr.12: 53307052-53307052 on Build GRCh38
rs148585651 Chr.12: 53306997-53306997 on Build GRCh38
rs778745212 Chr.12: 53307002-53307002 on Build GRCh38
rs1534282 Chr.12: 53307064-53307064 on Build GRCh38
rs748594633 Chr.12: 53307120-53307120 on Build GRCh38

View Full Product Details