Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96135218-96135489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASTL
Entrez Gene ID
431705
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Gene Aliases
SAS1B
UniGene
Hs.447993
Gene Chromosome Location
Chr.2: 96122817-96143059 on Build GRCh38
Transcripts
NM_001002036.3 XM_011511205.2 XM_011511206.2 XM_011511207.2 XM_011511208.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279009 c.196G>T p.E66* Chr.2: 96135398-96135398 on Build GRCh38
4096348 c.215T>G p.F72C Chr.2: 96135379-96135379 on Build GRCh38
166445 c.184C>T p.L62F Chr.2: 96135410-96135410 on Build GRCh38
6092774 c.216C>G p.F72L Chr.2: 96135378-96135378 on Build GRCh38
2825278 c.239G>A p.R80Q Chr.2: 96135355-96135355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750696859 Chr.2: 96135312-96135312 on Build GRCh38
rs373428105 Chr.2: 96135330-96135330 on Build GRCh38
rs777971926 Chr.2: 96135332-96135332 on Build GRCh38
rs755067196 Chr.2: 96135445-96135446 on Build GRCh38
rs745387575 Chr.2: 96135345-96135345 on Build GRCh38
rs897275531 Chr.2: 96135296-96135296 on Build GRCh38
rs377328988 Chr.2: 96135333-96135333 on Build GRCh38
rs771947479 Chr.2: 96135463-96135463 on Build GRCh38
rs986926239 Chr.2: 96135249-96135249 on Build GRCh38
rs890371027 Chr.2: 96135313-96135313 on Build GRCh38
rs184519754 Chr.2: 96135359-96135359 on Build GRCh38
rs374647937 Chr.2: 96135316-96135316 on Build GRCh38
rs1009427119 Chr.2: 96135348-96135348 on Build GRCh38
rs1029707238 Chr.2: 96135308-96135308 on Build GRCh38
rs770559800 Chr.2: 96135337-96135337 on Build GRCh38
rs775232913 Chr.2: 96135301-96135301 on Build GRCh38
rs368731223 Chr.2: 96135302-96135302 on Build GRCh38
rs375074901 Chr.2: 96135446-96135446 on Build GRCh38
rs370555704 Chr.2: 96135385-96135385 on Build GRCh38
rs908718519 Chr.2: 96135263-96135263 on Build GRCh38
rs143949503 Chr.2: 96135257-96135257 on Build GRCh38
rs368373087 Chr.2: 96135456-96135456 on Build GRCh38
rs775216219 Chr.2: 96135452-96135452 on Build GRCh38
rs754058897 Chr.2: 96135366-96135366 on Build GRCh38
rs755038635 Chr.2: 96135322-96135322 on Build GRCh38
rs532123683 Chr.2: 96135382-96135382 on Build GRCh38
rs997250772 Chr.2: 96135364-96135365 on Build GRCh38
rs199796644 Chr.2: 96135321-96135321 on Build GRCh38
rs769384491 Chr.2: 96135432-96135432 on Build GRCh38
rs1037091623 Chr.2: 96135251-96135251 on Build GRCh38
rs144520713 Chr.2: 96135411-96135411 on Build GRCh38
rs765193573 Chr.2: 96135371-96135371 on Build GRCh38
rs745520994 Chr.2: 96135362-96135362 on Build GRCh38
rs771568000 Chr.2: 96135369-96135369 on Build GRCh38
rs373382016 Chr.2: 96135355-96135355 on Build GRCh38
rs775529258 Chr.2: 96135352-96135352 on Build GRCh38
rs751917529 Chr.2: 96135401-96135401 on Build GRCh38
rs747053303 Chr.2: 96135422-96135424 on Build GRCh38
rs920379586 Chr.2: 96135309-96135309 on Build GRCh38
rs908684384 Chr.2: 96135434-96135434 on Build GRCh38
rs940392812 Chr.2: 96135287-96135287 on Build GRCh38
rs753291024 Chr.2: 96135372-96135372 on Build GRCh38
rs780844695 Chr.2: 96135416-96135416 on Build GRCh38
rs372369362 Chr.2: 96135310-96135310 on Build GRCh38
rs779491629 Chr.2: 96135326-96135326 on Build GRCh38
rs1037228605 Chr.2: 96135421-96135421 on Build GRCh38
rs774001295 Chr.2: 96135358-96135358 on Build GRCh38
rs370298467 Chr.2: 96135351-96135351 on Build GRCh38
rs745481743 Chr.2: 96135431-96135431 on Build GRCh38
rs910149540 Chr.2: 96135381-96135381 on Build GRCh38
rs776190309 Chr.2: 96135338-96135338 on Build GRCh38
rs780471358 Chr.2: 96135372-96135373 on Build GRCh38
rs377535345 Chr.2: 96135356-96135356 on Build GRCh38
rs748559131 Chr.2: 96135329-96135329 on Build GRCh38
rs375586097 Chr.2: 96135320-96135320 on Build GRCh38
rs904468244 Chr.2: 96135469-96135469 on Build GRCh38
rs777029980 Chr.2: 96135464-96135464 on Build GRCh38

View Full Product Details