Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137225495-137225750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYSRT1
Entrez Gene ID
375791
Gene Name
cysteine rich tail 1
Gene Aliases
C9orf169
UniGene
Hs.495540
Gene Chromosome Location
Chr.9: 137224635-137226311 on Build GRCh38
Transcripts
NM_199001.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7278372 c.68G>A p.R23Q Chr.9: 137225689-137225689 on Build GRCh38
1107079 c.27G>T p.K9N Chr.9: 137225648-137225648 on Build GRCh38
4791265 c.74A>G p.D25G Chr.9: 137225695-137225695 on Build GRCh38
3664248 c.17T>A p.M6K Chr.9: 137225638-137225638 on Build GRCh38
216131 c.22G>A p.V8I Chr.9: 137225643-137225643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753085142 Chr.9: 137225581-137225581 on Build GRCh38
rs763744130 Chr.9: 137225602-137225602 on Build GRCh38
rs778750467 Chr.9: 137225574-137225574 on Build GRCh38
rs922374903 Chr.9: 137225672-137225672 on Build GRCh38
rs545480758 Chr.9: 137225689-137225689 on Build GRCh38
rs908068526 Chr.9: 137225719-137225719 on Build GRCh38
rs990654678 Chr.9: 137225656-137225656 on Build GRCh38
rs138727583 Chr.9: 137225555-137225555 on Build GRCh38
rs762536612 Chr.9: 137225590-137225590 on Build GRCh38
rs768637587 Chr.9: 137225707-137225707 on Build GRCh38
rs573864886 Chr.9: 137225627-137225627 on Build GRCh38
rs1031694352 Chr.9: 137225618-137225618 on Build GRCh38
rs956087117 Chr.9: 137225648-137225648 on Build GRCh38
rs939743872 Chr.9: 137225731-137225731 on Build GRCh38
rs994614719 Chr.9: 137225524-137225538 on Build GRCh38
rs1017258456 Chr.9: 137225518-137225518 on Build GRCh38
rs200002597 Chr.9: 137225659-137225659 on Build GRCh38
rs1028659648 Chr.9: 137225557-137225557 on Build GRCh38
rs957586625 Chr.9: 137225623-137225623 on Build GRCh38
rs777842670 Chr.9: 137225715-137225715 on Build GRCh38
rs1038016579 Chr.9: 137225547-137225547 on Build GRCh38
rs749096383 Chr.9: 137225700-137225700 on Build GRCh38
rs762371960 Chr.9: 137225591-137225592 on Build GRCh38
rs751294443 Chr.9: 137225621-137225621 on Build GRCh38
rs866004302 Chr.9: 137225698-137225698 on Build GRCh38
rs542585171 Chr.9: 137225643-137225643 on Build GRCh38
rs573090509 Chr.9: 137225677-137225677 on Build GRCh38
rs955187668 Chr.9: 137225593-137225593 on Build GRCh38
rs1012713608 Chr.9: 137225527-137225527 on Build GRCh38
rs1047908306 Chr.9: 137225708-137225708 on Build GRCh38
rs910792659 Chr.9: 137225642-137225642 on Build GRCh38
rs577171158 Chr.9: 137225548-137225548 on Build GRCh38
rs980378123 Chr.9: 137225688-137225688 on Build GRCh38
rs756138128 Chr.9: 137225671-137225671 on Build GRCh38
rs768018887 Chr.9: 137225601-137225601 on Build GRCh38
rs557440965 Chr.9: 137225545-137225545 on Build GRCh38
rs998873658 Chr.9: 137225558-137225558 on Build GRCh38
rs779935067 Chr.9: 137225690-137225690 on Build GRCh38
rs556720856 Chr.9: 137225572-137225572 on Build GRCh38

View Full Product Details