Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 4222003-4222242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRA1D
Entrez Gene ID
146
Gene Name
adrenoceptor alpha 1D
Gene Aliases
ADRA1, ADRA1A, ADRA1R, ALPHA1, DAR, dJ779E11.2
UniGene
Hs.557
Gene Chromosome Location
Chr.20: 4220631-4249012 on Build GRCh38
Transcripts
NM_000678.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3746564 c.1142A>G p.E381G Chr.20: 4222100-4222100 on Build GRCh38
293280 c.1209_1210insC p.C404fs*>170 Chr.20: 4222032-4222033 on Build GRCh38
7218876 c.1205A>C p.Y402S Chr.20: 4222037-4222037 on Build GRCh38
1411893 c.1127A>G p.Q376R Chr.20: 4222115-4222115 on Build GRCh38
6840000 c.1166G>C p.W389S Chr.20: 4222076-4222076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772947925 Chr.20: 4222023-4222023 on Build GRCh38
rs761888252 Chr.20: 4222161-4222161 on Build GRCh38
rs767014721 Chr.20: 4222088-4222088 on Build GRCh38
rs1033870442 Chr.20: 4222213-4222213 on Build GRCh38
rs1001674314 Chr.20: 4222201-4222201 on Build GRCh38
rs773479185 Chr.20: 4222172-4222172 on Build GRCh38
rs765673280 Chr.20: 4222176-4222176 on Build GRCh38
rs767741828 Chr.20: 4222056-4222056 on Build GRCh38
rs61759834 Chr.20: 4222178-4222178 on Build GRCh38
rs766638344 Chr.20: 4222080-4222080 on Build GRCh38
rs978060380 Chr.20: 4222112-4222112 on Build GRCh38
rs755317160 Chr.20: 4222175-4222175 on Build GRCh38
rs906981583 Chr.20: 4222195-4222195 on Build GRCh38
rs915249753 Chr.20: 4222099-4222099 on Build GRCh38
rs745639541 Chr.20: 4222148-4222148 on Build GRCh38
rs835873 Chr.20: 4222155-4222155 on Build GRCh38
rs746650572 Chr.20: 4222132-4222132 on Build GRCh38
rs753185767 Chr.20: 4222177-4222177 on Build GRCh38
rs766806460 Chr.20: 4222174-4222174 on Build GRCh38
rs750578377 Chr.20: 4222063-4222063 on Build GRCh38
rs190503856 Chr.20: 4222134-4222134 on Build GRCh38
rs1038888440 Chr.20: 4222140-4222140 on Build GRCh38
rs776264034 Chr.20: 4222141-4222141 on Build GRCh38
rs970974390 Chr.20: 4222187-4222187 on Build GRCh38
rs753904294 Chr.20: 4222181-4222182 on Build GRCh38
rs776690942 Chr.20: 4222039-4222039 on Build GRCh38
rs761832897 Chr.20: 4222053-4222053 on Build GRCh38
rs58789310 Chr.20: 4222218-4222218 on Build GRCh38
rs372972092 Chr.20: 4222129-4222129 on Build GRCh38
rs779432485 Chr.20: 4222118-4222118 on Build GRCh38
rs1023970746 Chr.20: 4222185-4222185 on Build GRCh38
rs764277427 Chr.20: 4222046-4222046 on Build GRCh38
rs756536219 Chr.20: 4222077-4222077 on Build GRCh38
rs770847822 Chr.20: 4222026-4222026 on Build GRCh38
rs746810322 Chr.20: 4222024-4222024 on Build GRCh38
rs375905085 Chr.20: 4222150-4222150 on Build GRCh38
rs754074685 Chr.20: 4222130-4222130 on Build GRCh38
rs990115193 Chr.20: 4222096-4222096 on Build GRCh38
rs558211752 Chr.20: 4222214-4222214 on Build GRCh38
rs767582175 Chr.20: 4222162-4222162 on Build GRCh38
rs755408253 Chr.20: 4222093-4222093 on Build GRCh38
rs945340051 Chr.20: 4222103-4222103 on Build GRCh38
rs774326489 Chr.20: 4222048-4222048 on Build GRCh38
rs759373684 Chr.20: 4222041-4222041 on Build GRCh38
rs754417474 Chr.20: 4222081-4222081 on Build GRCh38

View Full Product Details