Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 42910681-42910954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf25
Entrez Gene ID
79020
Gene Name
chromosome 7 open reading frame 25
Gene Aliases
-
UniGene
Hs.698120
Gene Chromosome Location
Chr.7: 42909273-42913161 on Build GRCh38
Transcripts
NM_001099858.1 NM_024054.2 XM_005249844.3 XM_017012596.1 XM_017012597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7261840 c.228C>T p.I76I Chr.7: 42910847-42910847 on Build GRCh38
7275591 c.219G>T p.K73N Chr.7: 42910856-42910856 on Build GRCh38
6616553 c.202C>T p.R68* Chr.7: 42910873-42910873 on Build GRCh38
274316 c.83G>T p.R28I Chr.7: 42910818-42910818 on Build GRCh38
5000902 c.154G>T p.V52L Chr.7: 42910747-42910747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561007076 Chr.7: 42910709-42910709 on Build GRCh38
rs755248932 Chr.7: 42910890-42910890 on Build GRCh38
rs759222929 Chr.7: 42910823-42910823 on Build GRCh38
rs773139571 Chr.7: 42910797-42910797 on Build GRCh38
rs370667095 Chr.7: 42910835-42910835 on Build GRCh38
rs776694292 Chr.7: 42910892-42910892 on Build GRCh38
rs749639275 Chr.7: 42910887-42910887 on Build GRCh38
rs577661285 Chr.7: 42910925-42910925 on Build GRCh38
rs757801972 Chr.7: 42910722-42910722 on Build GRCh38
rs776384902 Chr.7: 42910821-42910821 on Build GRCh38
rs375214142 Chr.7: 42910838-42910838 on Build GRCh38
rs780614630 Chr.7: 42910868-42910868 on Build GRCh38
rs748250769 Chr.7: 42910904-42910904 on Build GRCh38
rs764970373 Chr.7: 42910831-42910831 on Build GRCh38
rs373871224 Chr.7: 42910793-42910793 on Build GRCh38
rs760598762 Chr.7: 42910805-42910805 on Build GRCh38
rs751039860 Chr.7: 42910860-42910860 on Build GRCh38
rs892886461 Chr.7: 42910777-42910777 on Build GRCh38
rs752224335 Chr.7: 42910718-42910718 on Build GRCh38
rs567722263 Chr.7: 42910779-42910779 on Build GRCh38
rs756677610 Chr.7: 42910866-42910866 on Build GRCh38
rs748282348 Chr.7: 42910784-42910784 on Build GRCh38
rs749578747 Chr.7: 42910754-42910754 on Build GRCh38
rs778987127 Chr.7: 42910910-42910912 on Build GRCh38
rs746315700 Chr.7: 42910732-42910732 on Build GRCh38
rs780559307 Chr.7: 42910751-42910751 on Build GRCh38
rs770285367 Chr.7: 42910747-42910747 on Build GRCh38
rs746994761 Chr.7: 42910927-42910927 on Build GRCh38
rs113806989 Chr.7: 42910880-42910880 on Build GRCh38
rs781731903 Chr.7: 42910723-42910723 on Build GRCh38
rs755020283 Chr.7: 42910876-42910877 on Build GRCh38
rs756490151 Chr.7: 42910719-42910719 on Build GRCh38
rs757943239 Chr.7: 42910847-42910847 on Build GRCh38
rs763448095 Chr.7: 42910856-42910856 on Build GRCh38
rs886433012 Chr.7: 42910818-42910818 on Build GRCh38
rs778340201 Chr.7: 42910715-42910715 on Build GRCh38
rs774334680 Chr.7: 42910766-42910766 on Build GRCh38
rs772259495 Chr.7: 42910919-42910919 on Build GRCh38
rs779208603 Chr.7: 42910900-42910900 on Build GRCh38
rs181507774 Chr.7: 42910845-42910845 on Build GRCh38
rs368085010 Chr.7: 42910807-42910807 on Build GRCh38

View Full Product Details