Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125823555-125823782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCCIP
Entrez Gene ID
56647
Gene Name
BRCA2 and CDKN1A interacting protein
Gene Aliases
TOK-1, TOK1
UniGene
Hs.370292
Gene Chromosome Location
Chr.10: 125823535-125853695 on Build GRCh38
Transcripts
NM_016567.3 NM_078468.2 NM_078469.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1197707 c.160G>A p.D54N Chr.10: 125823717-125823717 on Build GRCh38
1197708 c.160G>A p.D54N Chr.10: 125823717-125823717 on Build GRCh38
6128746 c.137A>G p.K46R Chr.10: 125823694-125823694 on Build GRCh38
6128748 c.137A>G p.K46R Chr.10: 125823694-125823694 on Build GRCh38
5567011 c.24T>C p.R8R Chr.10: 125823581-125823581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768971948 Chr.10: 125823651-125823651 on Build GRCh38
rs764622803 Chr.10: 125823744-125823744 on Build GRCh38
rs1021934496 Chr.10: 125823597-125823597 on Build GRCh38
rs185984858 Chr.10: 125823714-125823714 on Build GRCh38
rs757770433 Chr.10: 125823675-125823675 on Build GRCh38
rs142892325 Chr.10: 125823583-125823583 on Build GRCh38
rs975546277 Chr.10: 125823701-125823701 on Build GRCh38
rs761237671 Chr.10: 125823755-125823755 on Build GRCh38
rs758229366 Chr.10: 125823689-125823689 on Build GRCh38
rs779784228 Chr.10: 125823614-125823614 on Build GRCh38
rs776238100 Chr.10: 125823759-125823759 on Build GRCh38
rs918241440 Chr.10: 125823652-125823652 on Build GRCh38
rs141964850 Chr.10: 125823722-125823722 on Build GRCh38
rs772091914 Chr.10: 125823580-125823580 on Build GRCh38
rs775616562 Chr.10: 125823639-125823639 on Build GRCh38
rs368576919 Chr.10: 125823688-125823688 on Build GRCh38
rs746946148 Chr.10: 125823704-125823704 on Build GRCh38
rs749903009 Chr.10: 125823611-125823611 on Build GRCh38
rs774538161 Chr.10: 125823653-125823653 on Build GRCh38
rs199538471 Chr.10: 125823663-125823663 on Build GRCh38
rs753533068 Chr.10: 125823615-125823615 on Build GRCh38
rs942890141 Chr.10: 125823685-125823685 on Build GRCh38
rs144660023 Chr.10: 125823723-125823723 on Build GRCh38
rs372250278 Chr.10: 125823705-125823705 on Build GRCh38
rs776546665 Chr.10: 125823582-125823582 on Build GRCh38
rs772479642 Chr.10: 125823745-125823745 on Build GRCh38
rs200337015 Chr.10: 125823577-125823577 on Build GRCh38
rs773767862 Chr.10: 125823748-125823748 on Build GRCh38
rs764975377 Chr.10: 125823687-125823687 on Build GRCh38
rs200698402 Chr.10: 125823645-125823645 on Build GRCh38
rs773858139 Chr.10: 125823665-125823665 on Build GRCh38
rs113227232 Chr.10: 125823610-125823610 on Build GRCh38
rs755697021 Chr.10: 125823613-125823613 on Build GRCh38
rs368459243 Chr.10: 125823715-125823715 on Build GRCh38
rs878902048 Chr.10: 125823602-125823602 on Build GRCh38
rs775706553 Chr.10: 125823594-125823594 on Build GRCh38
rs550952791 Chr.10: 125823620-125823620 on Build GRCh38
rs769868910 Chr.10: 125823638-125823638 on Build GRCh38
rs1034132209 Chr.10: 125823637-125823637 on Build GRCh38
rs138579130 Chr.10: 125823576-125823576 on Build GRCh38
rs764363655 Chr.10: 125823596-125823596 on Build GRCh38
rs182757952 Chr.10: 125823695-125823695 on Build GRCh38
rs765972073 Chr.10: 125823672-125823672 on Build GRCh38
rs951090224 Chr.10: 125823662-125823662 on Build GRCh38
rs774646127 Chr.10: 125823742-125823742 on Build GRCh38
rs1010143629 Chr.10: 125823589-125823589 on Build GRCh38
rs992515101 Chr.10: 125823647-125823647 on Build GRCh38
rs761327886 Chr.10: 125823667-125823667 on Build GRCh38
rs138840348 Chr.10: 125823633-125823633 on Build GRCh38
rs758233007 Chr.10: 125823630-125823630 on Build GRCh38
rs200404406 Chr.10: 125823584-125823584 on Build GRCh38
rs752019800 Chr.10: 125823601-125823601 on Build GRCh38

View Full Product Details