Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206099863-206099616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf186
Entrez Gene ID
440712
Gene Name
chromosome 1 open reading frame 186
Gene Aliases
RHEX
UniGene
Hs.662248
Gene Chromosome Location
Chr.1: 206052723-206102701 on Build GRCh38
Transcripts
NM_001007544.3 XM_005272738.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1320543 c.153G>C p.Q51H Chr.1: 206099695-206099695 on Build GRCh38
1560311 c.256C>T p.H86Y Chr.1: 206099798-206099798 on Build GRCh38
3482379 c.255G>A p.R85R Chr.1: 206099797-206099797 on Build GRCh38
3482380 c.239C>T p.S80F Chr.1: 206099781-206099781 on Build GRCh38
5379390 c.157C>T p.P53S Chr.1: 206099699-206099699 on Build GRCh38
View More

View Full Product Details