Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48041400-48041671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP5
Entrez Gene ID
56344
Gene Name
calcium binding protein 5
Gene Aliases
CABP3
UniGene
Hs.117694
Gene Chromosome Location
Chr.19: 48029383-48048342 on Build GRCh38
Transcripts
NM_019855.4 XM_017026992.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7285107 c.94+10T>A p.? Chr.19: 48041563-48041563 on Build GRCh38
5390215 c.77G>A p.G26E Chr.19: 48041590-48041590 on Build GRCh38
6368585 c.82G>T p.D28Y Chr.19: 48041585-48041585 on Build GRCh38
3756994 c.87G>A p.E29E Chr.19: 48041580-48041580 on Build GRCh38
5910991 c.64-3C>T p.? Chr.19: 48041606-48041606 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771739690 Chr.19: 48041590-48041590 on Build GRCh38
rs768083218 Chr.19: 48041543-48041543 on Build GRCh38
rs377365861 Chr.19: 48041573-48041573 on Build GRCh38
rs748571516 Chr.19: 48041567-48041567 on Build GRCh38
rs571981679 Chr.19: 48041499-48041499 on Build GRCh38
rs765385834 Chr.19: 48041606-48041606 on Build GRCh38
rs1040097193 Chr.19: 48041506-48041506 on Build GRCh38
rs760491853 Chr.19: 48041592-48041592 on Build GRCh38
rs532159373 Chr.19: 48041444-48041444 on Build GRCh38
rs888074148 Chr.19: 48041622-48041636 on Build GRCh38
rs755392379 Chr.19: 48041565-48041565 on Build GRCh38
rs751908264 Chr.19: 48041561-48041561 on Build GRCh38
rs1038414155 Chr.19: 48041487-48041487 on Build GRCh38
rs773371492 Chr.19: 48041642-48041642 on Build GRCh38
rs35535557 Chr.19: 48041580-48041580 on Build GRCh38
rs201805500 Chr.19: 48041581-48041581 on Build GRCh38
rs375425951 Chr.19: 48041544-48041544 on Build GRCh38
rs764715142 Chr.19: 48041530-48041530 on Build GRCh38
rs745718288 Chr.19: 48041585-48041585 on Build GRCh38
rs567793963 Chr.19: 48041564-48041564 on Build GRCh38
rs941021300 Chr.19: 48041479-48041479 on Build GRCh38
rs763273974 Chr.19: 48041643-48041643 on Build GRCh38
rs888746881 Chr.19: 48041481-48041481 on Build GRCh38
rs1002489059 Chr.19: 48041521-48041521 on Build GRCh38
rs781752540 Chr.19: 48041566-48041566 on Build GRCh38
rs116560311 Chr.19: 48041583-48041583 on Build GRCh38
rs202039222 Chr.19: 48041624-48041624 on Build GRCh38
rs761999062 Chr.19: 48041523-48041523 on Build GRCh38
rs3745745 Chr.19: 48041496-48041496 on Build GRCh38
rs1048714356 Chr.19: 48041453-48041453 on Build GRCh38
rs534266476 Chr.19: 48041510-48041510 on Build GRCh38
rs369967045 Chr.19: 48041557-48041557 on Build GRCh38
rs554446379 Chr.19: 48041520-48041520 on Build GRCh38

View Full Product Details