Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23700687-23700958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM221A
Entrez Gene ID
340277
Gene Name
family with sequence similarity 221 member A
Gene Aliases
C7orf46
UniGene
Hs.592178
Gene Chromosome Location
Chr.7: 23680114-23703249 on Build GRCh38
Transcripts
NM_001127364.2 NM_001127365.2 NM_001300932.1 NM_199136.4 XM_011515369.2 XM_011515370.2 XM_011515371.2 XM_011515372.2 XM_017012134.1 XM_017012135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659530 c.766T>C p.S256P Chr.7: 23700806-23700806 on Build GRCh38
3637531 c.820C>A p.Q274K Chr.7: 23700860-23700860 on Build GRCh38
3637530 c.788A>C p.E263A Chr.7: 23700828-23700828 on Build GRCh38
7414710 c.754A>G p.S252G Chr.7: 23700794-23700794 on Build GRCh38
1283840 c.808G>A p.E270K Chr.7: 23700848-23700848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993253159 Chr.7: 23700776-23700776 on Build GRCh38
rs765563901 Chr.7: 23700910-23700910 on Build GRCh38
rs375667500 Chr.7: 23700800-23700800 on Build GRCh38
rs779411183 Chr.7: 23700915-23700915 on Build GRCh38
rs1018630152 Chr.7: 23700820-23700820 on Build GRCh38
rs773444167 Chr.7: 23700855-23700855 on Build GRCh38
rs779348765 Chr.7: 23700837-23700837 on Build GRCh38
rs1012183275 Chr.7: 23700727-23700727 on Build GRCh38
rs755508304 Chr.7: 23700835-23700835 on Build GRCh38
rs751159056 Chr.7: 23700890-23700890 on Build GRCh38
rs772327032 Chr.7: 23700744-23700744 on Build GRCh38
rs1031666621 Chr.7: 23700857-23700857 on Build GRCh38
rs142712999 Chr.7: 23700821-23700821 on Build GRCh38
rs765206059 Chr.7: 23700881-23700881 on Build GRCh38
rs752869411 Chr.7: 23700914-23700914 on Build GRCh38
rs767226864 Chr.7: 23700892-23700892 on Build GRCh38
rs890209935 Chr.7: 23700793-23700793 on Build GRCh38
rs752244905 Chr.7: 23700794-23700794 on Build GRCh38
rs765125999 Chr.7: 23700792-23700792 on Build GRCh38
rs747257628 Chr.7: 23700862-23700862 on Build GRCh38
rs776901204 Chr.7: 23700768-23700769 on Build GRCh38
rs760480959 Chr.7: 23700768-23700768 on Build GRCh38
rs754191888 Chr.7: 23700902-23700902 on Build GRCh38
rs749697607 Chr.7: 23700829-23700829 on Build GRCh38
rs771327899 Chr.7: 23700868-23700868 on Build GRCh38
rs1007696948 Chr.7: 23700805-23700805 on Build GRCh38
rs766309341 Chr.7: 23700769-23700769 on Build GRCh38
rs867316869 Chr.7: 23700834-23700834 on Build GRCh38
rs759665815 Chr.7: 23700769-23700769 on Build GRCh38
rs138016346 Chr.7: 23700784-23700784 on Build GRCh38
rs748202874 Chr.7: 23700841-23700841 on Build GRCh38
rs755314079 Chr.7: 23700909-23700909 on Build GRCh38
rs369844984 Chr.7: 23700764-23700764 on Build GRCh38
rs776535515 Chr.7: 23700869-23700869 on Build GRCh38
rs759470501 Chr.7: 23700873-23700873 on Build GRCh38
rs753779332 Chr.7: 23700770-23700770 on Build GRCh38
rs10244532 Chr.7: 23700882-23700882 on Build GRCh38
rs370344967 Chr.7: 23700817-23700817 on Build GRCh38
rs761868745 Chr.7: 23700736-23700736 on Build GRCh38
rs757901893 Chr.7: 23700796-23700796 on Build GRCh38
rs187670482 Chr.7: 23700816-23700816 on Build GRCh38
rs894621564 Chr.7: 23700724-23700724 on Build GRCh38
rs917298015 Chr.7: 23700887-23700887 on Build GRCh38
rs763539894 Chr.7: 23700886-23700886 on Build GRCh38
rs760955783 Chr.7: 23700715-23700715 on Build GRCh38
rs146025349 Chr.7: 23700850-23700850 on Build GRCh38
rs980550767 Chr.7: 23700925-23700925 on Build GRCh38
rs1021699907 Chr.7: 23700734-23700734 on Build GRCh38

View Full Product Details