Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 94528083-94528344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASD1
Entrez Gene ID
64921
Gene Name
CAS1 domain containing 1
Gene Aliases
C7orf12, NBLA04196
UniGene
Hs.260041
Gene Chromosome Location
Chr.7: 94509858-94633465 on Build GRCh38
Transcripts
NM_022900.4 XM_006716092.3 XM_006716093.3 XM_011516495.2 XM_017012546.1 XM_017012547.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6582711 c.424G>A p.G142S Chr.7: 94528215-94528215 on Build GRCh38
6270976 c.440G>T p.C147F Chr.7: 94528231-94528231 on Build GRCh38
6404594 c.436C>A p.Q146K Chr.7: 94528227-94528227 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752802503 Chr.7: 94528261-94528261 on Build GRCh38
rs200497452 Chr.7: 94528281-94528281 on Build GRCh38
rs964943741 Chr.7: 94528217-94528217 on Build GRCh38
rs1026339038 Chr.7: 94528152-94528152 on Build GRCh38
rs760150216 Chr.7: 94528184-94528184 on Build GRCh38
rs368247868 Chr.7: 94528181-94528181 on Build GRCh38
rs752752993 Chr.7: 94528149-94528149 on Build GRCh38
rs764101241 Chr.7: 94528274-94528276 on Build GRCh38
rs560425276 Chr.7: 94528268-94528268 on Build GRCh38
rs974988729 Chr.7: 94528142-94528142 on Build GRCh38
rs572120623 Chr.7: 94528275-94528275 on Build GRCh38
rs896470852 Chr.7: 94528131-94528131 on Build GRCh38
rs779377513 Chr.7: 94528271-94528271 on Build GRCh38
rs544625159 Chr.7: 94528275-94528276 on Build GRCh38
rs751613754 Chr.7: 94528285-94528310 on Build GRCh38
rs780467864 Chr.7: 94528282-94528282 on Build GRCh38
rs923210671 Chr.7: 94528273-94528273 on Build GRCh38
rs759885071 Chr.7: 94528273-94528273 on Build GRCh38
rs201576828 Chr.7: 94528205-94528205 on Build GRCh38
rs200785605 Chr.7: 94528293-94528293 on Build GRCh38
rs756312313 Chr.7: 94528153-94528153 on Build GRCh38
rs896119352 Chr.7: 94528272-94528272 on Build GRCh38
rs199658239 Chr.7: 94528287-94528287 on Build GRCh38
rs398005478 Chr.7: 94528286-94528286 on Build GRCh38
rs766025090 Chr.7: 94528251-94528254 on Build GRCh38
rs776913330 Chr.7: 94528260-94528261 on Build GRCh38
rs57770781 Chr.7: 94528274-94528274 on Build GRCh38
rs764958730 Chr.7: 94528236-94528236 on Build GRCh38
rs374684899 Chr.7: 94528146-94528146 on Build GRCh38
rs761585954 Chr.7: 94528235-94528235 on Build GRCh38
rs762908815 Chr.7: 94528240-94528240 on Build GRCh38
rs150000138 Chr.7: 94528214-94528214 on Build GRCh38
rs746237439 Chr.7: 94528280-94528280 on Build GRCh38
rs370364007 Chr.7: 94528265-94528265 on Build GRCh38
rs749663816 Chr.7: 94528156-94528156 on Build GRCh38
rs546766758 Chr.7: 94528263-94528263 on Build GRCh38
rs779433776 Chr.7: 94528172-94528172 on Build GRCh38
rs772731095 Chr.7: 94528162-94528166 on Build GRCh38
rs758842742 Chr.7: 94528273-94528274 on Build GRCh38
rs202237133 Chr.7: 94528179-94528179 on Build GRCh38
rs766251640 Chr.7: 94528245-94528245 on Build GRCh38
rs760277196 Chr.7: 94528228-94528229 on Build GRCh38
rs778178163 Chr.7: 94528155-94528155 on Build GRCh38
rs561355641 Chr.7: 94528274-94528275 on Build GRCh38
rs768325942 Chr.7: 94528204-94528204 on Build GRCh38
rs552469790 Chr.7: 94528311-94528311 on Build GRCh38
rs776135983 Chr.7: 94528291-94528291 on Build GRCh38
rs202044183 Chr.7: 94528286-94528286 on Build GRCh38

View Full Product Details