Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7742922-7743171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD209
Entrez Gene ID
30835
Gene Name
CD209 molecule
Gene Aliases
CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1
UniGene
Hs.278694
Gene Chromosome Location
Chr.19: 7739993-7747609 on Build GRCh38
Transcripts
NM_001144893.1 NM_001144894.1 NM_001144895.1 NM_001144896.1 NM_001144897.1 NM_001144899.1 NM_021155.3 NR_026692.1 XM_005272472.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
127575 c.1095C>T p.N365N Chr.19: 7743159-7743159 on Build GRCh38
5973251 c.1048T>C p.F350L Chr.19: 7743134-7743134 on Build GRCh38
5391155 c.1180C>T p.P394S Chr.19: 7743074-7743074 on Build GRCh38
7160843 c.1080C>T p.C360C Chr.19: 7743102-7743102 on Build GRCh38
5391152 c.1187C>T p.P396L Chr.19: 7743067-7743067 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776823008 Chr.19: 7742995-7742995 on Build GRCh38
rs369077812 Chr.19: 7742997-7742997 on Build GRCh38
rs770034550 Chr.19: 7743113-7743113 on Build GRCh38
rs780548412 Chr.19: 7743049-7743049 on Build GRCh38
rs149318064 Chr.19: 7743150-7743150 on Build GRCh38
rs143531930 Chr.19: 7743060-7743060 on Build GRCh38
rs763504034 Chr.19: 7743051-7743051 on Build GRCh38
rs767201321 Chr.19: 7743024-7743024 on Build GRCh38
rs773931530 Chr.19: 7743054-7743054 on Build GRCh38
rs777564858 Chr.19: 7743036-7743036 on Build GRCh38
rs751584461 Chr.19: 7743079-7743079 on Build GRCh38
rs373806865 Chr.19: 7743042-7743042 on Build GRCh38
rs749672042 Chr.19: 7743118-7743118 on Build GRCh38
rs11465393 Chr.19: 7743110-7743110 on Build GRCh38
rs755992712 Chr.19: 7743028-7743028 on Build GRCh38
rs749723284 Chr.19: 7743145-7743145 on Build GRCh38
rs867459495 Chr.19: 7743026-7743026 on Build GRCh38
rs778830508 Chr.19: 7743076-7743076 on Build GRCh38
rs775055595 Chr.19: 7743129-7743129 on Build GRCh38
rs752457226 Chr.19: 7743059-7743059 on Build GRCh38
rs11465394 Chr.19: 7743069-7743069 on Build GRCh38
rs1006859713 Chr.19: 7743024-7743024 on Build GRCh38
rs763850707 Chr.19: 7743027-7743027 on Build GRCh38
rs767060536 Chr.19: 7743058-7743058 on Build GRCh38
rs771595634 Chr.19: 7743052-7743052 on Build GRCh38
rs528658540 Chr.19: 7742967-7742967 on Build GRCh38
rs567293520 Chr.19: 7743073-7743073 on Build GRCh38
rs528884293 Chr.19: 7742989-7742989 on Build GRCh38
rs1016026388 Chr.19: 7743064-7743064 on Build GRCh38
rs560294624 Chr.19: 7742945-7742945 on Build GRCh38
rs758960447 Chr.19: 7743134-7743134 on Build GRCh38
rs376636419 Chr.19: 7742999-7742999 on Build GRCh38
rs766028272 Chr.19: 7743011-7743011 on Build GRCh38
rs374114895 Chr.19: 7743049-7743049 on Build GRCh38
rs200104082 Chr.19: 7743009-7743009 on Build GRCh38
rs879741982 Chr.19: 7743082-7743082 on Build GRCh38
rs771506047 Chr.19: 7743120-7743120 on Build GRCh38
rs768949282 Chr.19: 7742992-7742992 on Build GRCh38
rs376163704 Chr.19: 7743010-7743010 on Build GRCh38
rs1043940944 Chr.19: 7742944-7742945 on Build GRCh38
rs192979112 Chr.19: 7743012-7743012 on Build GRCh38
rs1035454461 Chr.19: 7743065-7743065 on Build GRCh38
rs755143789 Chr.19: 7743089-7743089 on Build GRCh38
rs901036660 Chr.19: 7743034-7743034 on Build GRCh38
rs770126743 Chr.19: 7742995-7742998 on Build GRCh38
rs775655494 Chr.19: 7742991-7742991 on Build GRCh38
rs774981477 Chr.19: 7743147-7743147 on Build GRCh38
rs375473990 Chr.19: 7743114-7743114 on Build GRCh38
rs201684036 Chr.19: 7743050-7743050 on Build GRCh38
rs771648411 Chr.19: 7743143-7743143 on Build GRCh38
rs370333676 Chr.19: 7743043-7743043 on Build GRCh38
rs754637180 Chr.19: 7743021-7743021 on Build GRCh38
rs138694095 Chr.19: 7743094-7743094 on Build GRCh38

View Full Product Details