Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 81046008-81046241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP3
Entrez Gene ID
651
Gene Name
bone morphogenetic protein 3
Gene Aliases
BMP-3A
UniGene
Hs.387411
Gene Chromosome Location
Chr.4: 81030778-81057531 on Build GRCh38
Transcripts
NM_001201.2 XM_006714291.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5799786 c.690G>C p.K230N Chr.4: 81046111-81046111 on Build GRCh38
1694598 c.796C>T p.P266S Chr.4: 81046217-81046217 on Build GRCh38
5995706 c.632A>T p.E211V Chr.4: 81046053-81046053 on Build GRCh38
4772624 c.676C>T p.R226C Chr.4: 81046097-81046097 on Build GRCh38
1726838 c.691A>T p.R231W Chr.4: 81046112-81046112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769459824 Chr.4: 81046191-81046191 on Build GRCh38
rs528914843 Chr.4: 81046062-81046062 on Build GRCh38
rs746981136 Chr.4: 81046161-81046161 on Build GRCh38
rs1007409366 Chr.4: 81046162-81046162 on Build GRCh38
rs866813886 Chr.4: 81046217-81046217 on Build GRCh38
rs750540157 Chr.4: 81046041-81046041 on Build GRCh38
rs760788016 Chr.4: 81046131-81046131 on Build GRCh38
rs1037519829 Chr.4: 81046108-81046108 on Build GRCh38
rs748030422 Chr.4: 81046071-81046071 on Build GRCh38
rs965953329 Chr.4: 81046197-81046197 on Build GRCh38
rs951813712 Chr.4: 81046137-81046137 on Build GRCh38
rs754084275 Chr.4: 81046138-81046138 on Build GRCh38
rs145436929 Chr.4: 81046109-81046109 on Build GRCh38
rs370315521 Chr.4: 81046087-81046087 on Build GRCh38
rs1055877104 Chr.4: 81046036-81046036 on Build GRCh38
rs367604119 Chr.4: 81046113-81046113 on Build GRCh38
rs777732680 Chr.4: 81046156-81046156 on Build GRCh38
rs750935828 Chr.4: 81046116-81046116 on Build GRCh38
rs766425555 Chr.4: 81046133-81046133 on Build GRCh38
rs780292516 Chr.4: 81046053-81046053 on Build GRCh38
rs536817590 Chr.4: 81046145-81046145 on Build GRCh38
rs200302858 Chr.4: 81046208-81046208 on Build GRCh38
rs149231562 Chr.4: 81046195-81046195 on Build GRCh38
rs957904588 Chr.4: 81046178-81046178 on Build GRCh38
rs140292630 Chr.4: 81046163-81046163 on Build GRCh38
rs571119277 Chr.4: 81046139-81046139 on Build GRCh38
rs774714825 Chr.4: 81046103-81046103 on Build GRCh38
rs774659530 Chr.4: 81046194-81046194 on Build GRCh38
rs745469742 Chr.4: 81046184-81046184 on Build GRCh38
rs758369373 Chr.4: 81046149-81046149 on Build GRCh38
rs6831040 Chr.4: 81046034-81046034 on Build GRCh38
rs754806806 Chr.4: 81046064-81046064 on Build GRCh38
rs772145730 Chr.4: 81046084-81046084 on Build GRCh38
rs998297057 Chr.4: 81046123-81046123 on Build GRCh38
rs756251230 Chr.4: 81046047-81046047 on Build GRCh38
rs776336946 Chr.4: 81046097-81046097 on Build GRCh38
rs34505126 Chr.4: 81046086-81046086 on Build GRCh38
rs373752528 Chr.4: 81046098-81046098 on Build GRCh38
rs770871155 Chr.4: 81046147-81046147 on Build GRCh38
rs752059197 Chr.4: 81046038-81046038 on Build GRCh38
rs114620549 Chr.4: 81046085-81046085 on Build GRCh38
rs778916024 Chr.4: 81046069-81046069 on Build GRCh38
rs975961336 Chr.4: 81046209-81046209 on Build GRCh38
rs780576204 Chr.4: 81046182-81046182 on Build GRCh38

View Full Product Details