Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 95611235-95611474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH18A1
Entrez Gene ID
5832
Gene Name
aldehyde dehydrogenase 18 family member A1
Gene Aliases
ADCL3, ARCL3A, GSAS, P5CS, PYCS, SPG9A, SPG9B
UniGene
Hs.500645
Gene Chromosome Location
Chr.10: 95605929-95656810 on Build GRCh38
Transcripts
NM_001017423.1 NM_001323412.1 NM_001323413.1 NM_001323414.1 NM_001323415.1 NM_001323416.1 NM_001323417.1 NM_001323418.1 NM_001323419.1 NM_002860.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3738803 c.1977C>T p.S659S Chr.10: 95611389-95611389 on Build GRCh38
6598165 c.2109C>T p.D703D Chr.10: 95611257-95611257 on Build GRCh38
6244484 c.1992C>A p.L664L Chr.10: 95611374-95611374 on Build GRCh38
4977149 c.2105A>G p.E702G Chr.10: 95611261-95611261 on Build GRCh38
3442141 c.2008G>A p.D670N Chr.10: 95611358-95611358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78731297 Chr.10: 95611365-95611365 on Build GRCh38
rs979536943 Chr.10: 95611261-95611261 on Build GRCh38
rs1012303779 Chr.10: 95611321-95611321 on Build GRCh38
rs746723369 Chr.10: 95611265-95611265 on Build GRCh38
rs562987600 Chr.10: 95611383-95611383 on Build GRCh38
rs772496373 Chr.10: 95611294-95611294 on Build GRCh38
rs989031241 Chr.10: 95611342-95611342 on Build GRCh38
rs776999689 Chr.10: 95611431-95611431 on Build GRCh38
rs761988720 Chr.10: 95611358-95611358 on Build GRCh38
rs779227894 Chr.10: 95611395-95611395 on Build GRCh38
rs189509801 Chr.10: 95611392-95611392 on Build GRCh38
rs139742227 Chr.10: 95611296-95611296 on Build GRCh38
rs764233894 Chr.10: 95611325-95611325 on Build GRCh38
rs746442782 Chr.10: 95611411-95611411 on Build GRCh38
rs375705932 Chr.10: 95611268-95611268 on Build GRCh38
rs761744291 Chr.10: 95611435-95611435 on Build GRCh38
rs560801065 Chr.10: 95611300-95611300 on Build GRCh38
rs771292407 Chr.10: 95611278-95611278 on Build GRCh38
rs1804934 Chr.10: 95611389-95611389 on Build GRCh38
rs780844696 Chr.10: 95611387-95611387 on Build GRCh38
rs761041543 Chr.10: 95611322-95611322 on Build GRCh38
rs758116463 Chr.10: 95611367-95611367 on Build GRCh38
rs769976269 Chr.10: 95611436-95611436 on Build GRCh38
rs774952464 Chr.10: 95611287-95611287 on Build GRCh38
rs746150080 Chr.10: 95611288-95611288 on Build GRCh38
rs150980819 Chr.10: 95611363-95611363 on Build GRCh38
rs766264810 Chr.10: 95611372-95611372 on Build GRCh38
rs751181507 Chr.10: 95611373-95611373 on Build GRCh38
rs761417811 Chr.10: 95611453-95611453 on Build GRCh38
rs145197911 Chr.10: 95611420-95611420 on Build GRCh38
rs779516879 Chr.10: 95611257-95611257 on Build GRCh38
rs768964431 Chr.10: 95611424-95611424 on Build GRCh38
rs145806693 Chr.10: 95611326-95611326 on Build GRCh38
rs758219423 Chr.10: 95611256-95611256 on Build GRCh38
rs780721947 Chr.10: 95611269-95611269 on Build GRCh38
rs905973576 Chr.10: 95611406-95611406 on Build GRCh38
rs772316092 Chr.10: 95611413-95611413 on Build GRCh38

View Full Product Details