Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43229324-43229559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNPH1
Entrez Gene ID
10591
Gene Name
2'-deoxynucleoside 5'-phosphate N-hydrolase 1
Gene Aliases
C6orf108, RCL, dJ330M21.3
UniGene
Hs.109752
Gene Chromosome Location
Chr.6: 43225629-43229473 on Build GRCh38
Transcripts
NM_006443.2 NM_199184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4801530 c.42G>A p.E14E Chr.6: 43229415-43229415 on Build GRCh38
6385527 c.101A>G p.E34G Chr.6: 43229356-43229356 on Build GRCh38
7317213 c.31G>A p.E11K Chr.6: 43229426-43229426 on Build GRCh38
6497644 c.51G>A p.E17E Chr.6: 43229406-43229406 on Build GRCh38
5657230 c.15G>T p.M5I Chr.6: 43229442-43229442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746733806 Chr.6: 43229384-43229384 on Build GRCh38
rs933432507 Chr.6: 43229534-43229534 on Build GRCh38
rs1052985812 Chr.6: 43229412-43229412 on Build GRCh38
rs916481611 Chr.6: 43229405-43229405 on Build GRCh38
rs557206532 Chr.6: 43229498-43229498 on Build GRCh38
rs568605728 Chr.6: 43229473-43229473 on Build GRCh38
rs762528187 Chr.6: 43229411-43229411 on Build GRCh38
rs1026472444 Chr.6: 43229444-43229444 on Build GRCh38
rs769278085 Chr.6: 43229409-43229409 on Build GRCh38
rs2295790 Chr.6: 43229441-43229441 on Build GRCh38
rs1055091385 Chr.6: 43229393-43229393 on Build GRCh38
rs974792722 Chr.6: 43229479-43229479 on Build GRCh38
rs927702086 Chr.6: 43229522-43229522 on Build GRCh38
rs760924174 Chr.6: 43229416-43229416 on Build GRCh38
rs866717784 Chr.6: 43229378-43229378 on Build GRCh38
rs758532471 Chr.6: 43229439-43229439 on Build GRCh38
rs879437165 Chr.6: 43229491-43229491 on Build GRCh38
rs943379931 Chr.6: 43229475-43229475 on Build GRCh38
rs369710271 Chr.6: 43229415-43229415 on Build GRCh38
rs759253546 Chr.6: 43229408-43229408 on Build GRCh38
rs747011368 Chr.6: 43229446-43229446 on Build GRCh38
rs996577653 Chr.6: 43229454-43229454 on Build GRCh38
rs779283651 Chr.6: 43229434-43229434 on Build GRCh38
rs901876842 Chr.6: 43229438-43229438 on Build GRCh38
rs1035409317 Chr.6: 43229436-43229436 on Build GRCh38
rs770630065 Chr.6: 43229458-43229458 on Build GRCh38
rs781020654 Chr.6: 43229466-43229466 on Build GRCh38
rs537640808 Chr.6: 43229482-43229482 on Build GRCh38
rs766889690 Chr.6: 43229420-43229420 on Build GRCh38
rs966450475 Chr.6: 43229495-43229495 on Build GRCh38
rs957594128 Chr.6: 43229460-43229460 on Build GRCh38
rs770810122 Chr.6: 43229386-43229386 on Build GRCh38
rs777802179 Chr.6: 43229376-43229376 on Build GRCh38
rs866059058 Chr.6: 43229395-43229395 on Build GRCh38
rs776237012 Chr.6: 43229389-43229389 on Build GRCh38
rs13215758 Chr.6: 43229517-43229517 on Build GRCh38
rs992282412 Chr.6: 43229443-43229443 on Build GRCh38
rs780115160 Chr.6: 43229472-43229472 on Build GRCh38
rs748255178 Chr.6: 43229347-43229347 on Build GRCh38
rs905550108 Chr.6: 43229431-43229431 on Build GRCh38
rs913099858 Chr.6: 43229539-43229539 on Build GRCh38
rs1004930254 Chr.6: 43229432-43229432 on Build GRCh38
rs960430200 Chr.6: 43229538-43229538 on Build GRCh38
rs891716079 Chr.6: 43229413-43229413 on Build GRCh38
rs879865521 Chr.6: 43229485-43229486 on Build GRCh38
rs754105901 Chr.6: 43229422-43229422 on Build GRCh38
rs755195975 Chr.6: 43229424-43229424 on Build GRCh38
rs943446552 Chr.6: 43229377-43229377 on Build GRCh38
rs774921547 Chr.6: 43229410-43229410 on Build GRCh38
rs886545251 Chr.6: 43229419-43229419 on Build GRCh38
rs752785239 Chr.6: 43229437-43229437 on Build GRCh38
rs771842003 Chr.6: 43229351-43229351 on Build GRCh38

View Full Product Details