Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 428686-428939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC77
Entrez Gene ID
84318
Gene Name
coiled-coil domain containing 77
Gene Aliases
-
UniGene
Hs.631656
Gene Chromosome Location
Chr.12: 389350-442642 on Build GRCh38
Transcripts
NM_001130146.1 NM_001130147.1 NM_001130148.1 NM_032358.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5528281 c.452C>T p.A151V Chr.12: 428807-428807 on Build GRCh38
5970757 c.417G>A p.E139E Chr.12: 428772-428772 on Build GRCh38
416173 c.500C>T p.P167L Chr.12: 428855-428855 on Build GRCh38
4575550 c.452C>G p.A151G Chr.12: 428807-428807 on Build GRCh38
6137302 c.414G>T p.R138S Chr.12: 428769-428769 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781282189 Chr.12: 428889-428889 on Build GRCh38
rs369263756 Chr.12: 428726-428726 on Build GRCh38
rs376422753 Chr.12: 428878-428878 on Build GRCh38
rs929163487 Chr.12: 428833-428833 on Build GRCh38
rs768885093 Chr.12: 428882-428882 on Build GRCh38
rs776970210 Chr.12: 428733-428733 on Build GRCh38
rs769928115 Chr.12: 428784-428784 on Build GRCh38
rs777815645 Chr.12: 428864-428867 on Build GRCh38
rs376035524 Chr.12: 428871-428871 on Build GRCh38
rs200465914 Chr.12: 428880-428880 on Build GRCh38
rs984526282 Chr.12: 428861-428861 on Build GRCh38
rs1022837341 Chr.12: 428895-428895 on Build GRCh38
rs748614518 Chr.12: 428901-428901 on Build GRCh38
rs200870206 Chr.12: 428776-428776 on Build GRCh38
rs751119759 Chr.12: 428807-428807 on Build GRCh38
rs372464070 Chr.12: 428854-428854 on Build GRCh38
rs758755350 Chr.12: 428837-428837 on Build GRCh38
rs775783672 Chr.12: 428719-428719 on Build GRCh38
rs942098493 Chr.12: 428859-428859 on Build GRCh38
rs780397607 Chr.12: 428845-428845 on Build GRCh38
rs762338201 Chr.12: 428742-428742 on Build GRCh38
rs773566627 Chr.12: 428756-428756 on Build GRCh38
rs754453910 Chr.12: 428809-428809 on Build GRCh38
rs752600033 Chr.12: 428820-428820 on Build GRCh38
rs765540054 Chr.12: 428744-428744 on Build GRCh38
rs747122768 Chr.12: 428847-428847 on Build GRCh38
rs762597198 Chr.12: 428759-428759 on Build GRCh38
rs756023593 Chr.12: 428824-428824 on Build GRCh38
rs777436675 Chr.12: 428829-428829 on Build GRCh38
rs374707472 Chr.12: 428779-428779 on Build GRCh38
rs375742373 Chr.12: 428828-428828 on Build GRCh38
rs770268539 Chr.12: 428905-428905 on Build GRCh38
rs766975329 Chr.12: 428819-428819 on Build GRCh38
rs766015581 Chr.12: 428789-428789 on Build GRCh38
rs879861185 Chr.12: 428860-428860 on Build GRCh38
rs199659409 Chr.12: 428827-428827 on Build GRCh38
rs964988653 Chr.12: 428772-428772 on Build GRCh38
rs747067870 Chr.12: 428723-428723 on Build GRCh38
rs201844836 Chr.12: 428830-428830 on Build GRCh38
rs751037481 Chr.12: 428869-428869 on Build GRCh38

View Full Product Details