Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130005135-130005408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCORL1
Entrez Gene ID
63035
Gene Name
BCL6 corepressor-like 1
Gene Aliases
BCoR-L1, CXorf10
UniGene
Hs.496748
Gene Chromosome Location
Chr.X: 129980285-130058083 on Build GRCh38
Transcripts
NM_001184772.2 NM_021946.4 XM_005262452.4 XM_005262453.4 XM_005262454.3 XM_005262455.4 XM_005262456.4 XM_006724776.3 XM_006724777.3 XM_006724779.2 XM_017029721.1 XM_017029722.1

SNP Detail

SNP ID Chromosome Location
rs772758161 Chr.X: 130005220-130005220 on Build GRCh38
rs145654461 Chr.X: 130005244-130005244 on Build GRCh38
rs777018072 Chr.X: 130005355-130005355 on Build GRCh38
rs751755820 Chr.X: 130005256-130005256 on Build GRCh38
rs201118119 Chr.X: 130005262-130005262 on Build GRCh38
rs781631018 Chr.X: 130005333-130005340 on Build GRCh38
rs765554234 Chr.X: 130005239-130005239 on Build GRCh38
rs376483067 Chr.X: 130005369-130005369 on Build GRCh38
rs772700878 Chr.X: 130005345-130005345 on Build GRCh38
rs138189929 Chr.X: 130005292-130005292 on Build GRCh38
rs369214982 Chr.X: 130005218-130005218 on Build GRCh38
rs757653362 Chr.X: 130005330-130005330 on Build GRCh38
rs1057180410 Chr.X: 130005310-130005310 on Build GRCh38
rs749188963 Chr.X: 130005287-130005287 on Build GRCh38
rs759885411 Chr.X: 130005230-130005230 on Build GRCh38
rs769107488 Chr.X: 130005340-130005340 on Build GRCh38
rs747775616 Chr.X: 130005334-130005334 on Build GRCh38
rs906785824 Chr.X: 130005261-130005261 on Build GRCh38
rs761135980 Chr.X: 130005227-130005227 on Build GRCh38
rs758746997 Chr.X: 130005248-130005248 on Build GRCh38
rs777972699 Chr.X: 130005252-130005252 on Build GRCh38
rs746583844 Chr.X: 130005353-130005353 on Build GRCh38
rs779984923 Chr.X: 130005273-130005273 on Build GRCh38
rs778701486 Chr.X: 130005293-130005293 on Build GRCh38
rs770358152 Chr.X: 130005368-130005368 on Build GRCh38
rs754135968 Chr.X: 130005229-130005229 on Build GRCh38
rs373485840 Chr.X: 130005309-130005309 on Build GRCh38

View Full Product Details