Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201491939-201492172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALS2CR11
Entrez Gene ID
151254
Gene Name
amyotrophic lateral sclerosis 2 chromosome region candidate 11
Gene Aliases
-
UniGene
Hs.335788
Gene Chromosome Location
Chr.2: 201487421-201619182 on Build GRCh38
Transcripts
NM_001168216.1 NM_001168221.1 NM_152525.5 XM_006712331.3 XM_006712332.3 XM_006712333.3 XM_006712335.3 XM_006712336.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6543805 c.4338T>A p.S1446R Chr.2: 201492003-201492003 on Build GRCh38
4838760 c.4245G>T p.K1415N Chr.2: 201492096-201492096 on Build GRCh38
5437470 c.4244A>T p.K1415M Chr.2: 201492097-201492097 on Build GRCh38
1591741 c.4231C>T p.R1411* Chr.2: 201492110-201492110 on Build GRCh38
1153700 c.4321G>A p.E1441K Chr.2: 201492020-201492020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016533471 Chr.2: 201492116-201492116 on Build GRCh38
rs765033918 Chr.2: 201491996-201491996 on Build GRCh38
rs763955746 Chr.2: 201492048-201492048 on Build GRCh38
rs571028000 Chr.2: 201492110-201492110 on Build GRCh38
rs568868463 Chr.2: 201492114-201492114 on Build GRCh38
rs751454434 Chr.2: 201492081-201492081 on Build GRCh38
rs530754596 Chr.2: 201492020-201492020 on Build GRCh38
rs1004787425 Chr.2: 201492109-201492109 on Build GRCh38
rs187815911 Chr.2: 201492138-201492138 on Build GRCh38
rs775599842 Chr.2: 201492010-201492010 on Build GRCh38
rs75689388 Chr.2: 201492026-201492026 on Build GRCh38
rs557048083 Chr.2: 201492080-201492080 on Build GRCh38

View Full Product Details