Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36149485-36149738 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPNS1
Entrez Gene ID
826
Gene Name
calpain small subunit 1
Gene Aliases
CALPAIN4, CANP, CANPS, CAPN4, CDPS, CSS1
UniGene
Hs.515371
Gene Chromosome Location
Chr.19: 36139926-36150353 on Build GRCh38
Transcripts
NM_001003962.2 NM_001302632.1 NM_001302633.1 NM_001749.3 XM_005259295.1 XM_005259296.1 XM_011527359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6568146 c.762C>T p.I254I Chr.19: 36149618-36149618 on Build GRCh38
3532899 c.780+10C>G p.? Chr.19: 36149646-36149646 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs17879753 Chr.19: 36149664-36149664 on Build GRCh38
rs553595702 Chr.19: 36149573-36149573 on Build GRCh38
rs535283662 Chr.19: 36149523-36149523 on Build GRCh38
rs752226310 Chr.19: 36149646-36149646 on Build GRCh38
rs765107361 Chr.19: 36149641-36149641 on Build GRCh38
rs917979079 Chr.19: 36149689-36149689 on Build GRCh38
rs756919953 Chr.19: 36149661-36149661 on Build GRCh38
rs763794605 Chr.19: 36149649-36149649 on Build GRCh38
rs200436917 Chr.19: 36149667-36149667 on Build GRCh38
rs1013824519 Chr.19: 36149579-36149579 on Build GRCh38
rs113520816 Chr.19: 36149577-36149577 on Build GRCh38
rs776070253 Chr.19: 36149518-36149518 on Build GRCh38
rs770790595 Chr.19: 36149608-36149608 on Build GRCh38
rs773608674 Chr.19: 36149599-36149599 on Build GRCh38
rs779013240 Chr.19: 36149566-36149566 on Build GRCh38
rs756139280 Chr.19: 36149566-36149568 on Build GRCh38
rs759505309 Chr.19: 36149612-36149612 on Build GRCh38
rs376137879 Chr.19: 36149679-36149679 on Build GRCh38
rs931396263 Chr.19: 36149562-36149562 on Build GRCh38
rs751275273 Chr.19: 36149660-36149660 on Build GRCh38
rs192316761 Chr.19: 36149696-36149696 on Build GRCh38
rs369952446 Chr.19: 36149568-36149568 on Build GRCh38
rs560742467 Chr.19: 36149702-36149702 on Build GRCh38
rs749740689 Chr.19: 36149542-36149542 on Build GRCh38
rs769050033 Chr.19: 36149552-36149552 on Build GRCh38
rs114646517 Chr.19: 36149621-36149621 on Build GRCh38
rs780013833 Chr.19: 36149669-36149669 on Build GRCh38
rs762406499 Chr.19: 36149647-36149647 on Build GRCh38
rs919982261 Chr.19: 36149551-36149551 on Build GRCh38
rs760532259 Chr.19: 36149603-36149603 on Build GRCh38
rs1037519730 Chr.19: 36149549-36149549 on Build GRCh38
rs111468810 Chr.19: 36149665-36149665 on Build GRCh38
rs780529823 Chr.19: 36149540-36149540 on Build GRCh38

View Full Product Details