Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73311189-73311434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF17
Entrez Gene ID
9828
Gene Name
Rho guanine nucleotide exchange factor 17
Gene Aliases
P164RHOGEF, TEM4, p164-RhoGEF
UniGene
Hs.533719
Gene Chromosome Location
Chr.11: 73305961-73369380 on Build GRCh38
Transcripts
NM_014786.3 XM_017018623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6604053 c.2552G>A p.R851Q Chr.11: 73311190-73311190 on Build GRCh38
4146316 c.2567T>G p.M856R Chr.11: 73311205-73311205 on Build GRCh38
7284264 c.2609G>A p.R870Q Chr.11: 73311247-73311247 on Build GRCh38
3870179 c.2753T>C p.I918T Chr.11: 73311391-73311391 on Build GRCh38
931800 c.2604G>A p.E868E Chr.11: 73311242-73311242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112430027 Chr.11: 73311233-73311233 on Build GRCh38
rs920674049 Chr.11: 73311303-73311303 on Build GRCh38
rs750854808 Chr.11: 73311237-73311237 on Build GRCh38
rs772335178 Chr.11: 73311380-73311380 on Build GRCh38
rs757320208 Chr.11: 73311358-73311358 on Build GRCh38
rs201830165 Chr.11: 73311283-73311283 on Build GRCh38
rs758043385 Chr.11: 73311340-73311340 on Build GRCh38
rs903052982 Chr.11: 73311348-73311348 on Build GRCh38
rs375013444 Chr.11: 73311234-73311234 on Build GRCh38
rs533969393 Chr.11: 73311389-73311389 on Build GRCh38
rs751073477 Chr.11: 73311357-73311357 on Build GRCh38
rs957284488 Chr.11: 73311218-73311218 on Build GRCh38
rs746855834 Chr.11: 73311209-73311209 on Build GRCh38
rs766860579 Chr.11: 73311335-73311335 on Build GRCh38
rs772093395 Chr.11: 73311322-73311322 on Build GRCh38
rs754216768 Chr.11: 73311337-73311337 on Build GRCh38
rs372044743 Chr.11: 73311379-73311379 on Build GRCh38
rs770957933 Chr.11: 73311307-73311307 on Build GRCh38
rs781197480 Chr.11: 73311313-73311313 on Build GRCh38
rs777185020 Chr.11: 73311346-73311346 on Build GRCh38
rs746964996 Chr.11: 73311302-73311302 on Build GRCh38
rs764905511 Chr.11: 73311339-73311339 on Build GRCh38
rs762575728 Chr.11: 73311409-73311409 on Build GRCh38
rs781215981 Chr.11: 73311359-73311359 on Build GRCh38
rs759920014 Chr.11: 73311397-73311397 on Build GRCh38
rs765566202 Chr.11: 73311400-73311400 on Build GRCh38
rs903203449 Chr.11: 73311270-73311270 on Build GRCh38
rs761982935 Chr.11: 73311320-73311320 on Build GRCh38
rs771434898 Chr.11: 73311393-73311393 on Build GRCh38
rs765506319 Chr.11: 73311344-73311344 on Build GRCh38
rs756622785 Chr.11: 73311244-73311244 on Build GRCh38
rs930668153 Chr.11: 73311312-73311312 on Build GRCh38
rs777784126 Chr.11: 73311301-73311301 on Build GRCh38
rs754198385 Chr.11: 73311258-73311258 on Build GRCh38
rs377082578 Chr.11: 73311211-73311211 on Build GRCh38
rs369489970 Chr.11: 73311282-73311282 on Build GRCh38
rs760691466 Chr.11: 73311383-73311383 on Build GRCh38
rs777086331 Chr.11: 73311396-73311396 on Build GRCh38
rs554470373 Chr.11: 73311361-73311361 on Build GRCh38
rs773311250 Chr.11: 73311323-73311323 on Build GRCh38
rs892897422 Chr.11: 73311394-73311394 on Build GRCh38
rs763303354 Chr.11: 73311221-73311221 on Build GRCh38
rs769011649 Chr.11: 73311317-73311317 on Build GRCh38
rs774545723 Chr.11: 73311231-73311231 on Build GRCh38
rs376703409 Chr.11: 73311225-73311225 on Build GRCh38
rs745642279 Chr.11: 73311316-73311316 on Build GRCh38
rs764403659 Chr.11: 73311224-73311224 on Build GRCh38
rs753070536 Chr.11: 73311408-73311408 on Build GRCh38
rs769672128 Chr.11: 73311362-73311362 on Build GRCh38
rs910174113 Chr.11: 73311372-73311372 on Build GRCh38
rs139353082 Chr.11: 73311325-73311325 on Build GRCh38
rs369906561 Chr.11: 73311411-73311411 on Build GRCh38
rs370660401 Chr.11: 73311247-73311247 on Build GRCh38
rs141908042 Chr.11: 73311382-73311382 on Build GRCh38
rs776541626 Chr.11: 73311215-73311215 on Build GRCh38
rs569021852 Chr.11: 73311248-73311248 on Build GRCh38
rs779218653 Chr.11: 73311365-73311365 on Build GRCh38
rs774369382 Chr.11: 73311319-73311319 on Build GRCh38
rs759830696 Chr.11: 73311338-73311338 on Build GRCh38
rs146893545 Chr.11: 73311298-73311298 on Build GRCh38
rs751125685 Chr.11: 73311412-73311412 on Build GRCh38
rs754685199 Chr.11: 73311268-73311268 on Build GRCh38

View Full Product Details