Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 130821233-130821504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY8
Entrez Gene ID
114
Gene Name
adenylate cyclase 8
Gene Aliases
AC8, ADCY3, HBAC1
UniGene
Hs.591859
Gene Chromosome Location
Chr.8: 130780300-131041604 on Build GRCh38
Transcripts
NM_001115.2 XM_005250769.3 XM_006716501.3 XM_017013006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
308946 c.2745T>C p.H915H Chr.8: 130821351-130821351 on Build GRCh38
7354554 c.2713A>T p.M905L Chr.8: 130821383-130821383 on Build GRCh38
3663563 c.2734G>T p.V912L Chr.8: 130821362-130821362 on Build GRCh38
6052520 c.2688G>T p.G896G Chr.8: 130821408-130821408 on Build GRCh38
6179983 c.2676-2A>G p.? Chr.8: 130821422-130821422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965819161 Chr.8: 130821337-130821337 on Build GRCh38
rs769049662 Chr.8: 130821335-130821335 on Build GRCh38
rs769988069 Chr.8: 130821459-130821459 on Build GRCh38
rs374458875 Chr.8: 130821302-130821302 on Build GRCh38
rs369816294 Chr.8: 130821326-130821326 on Build GRCh38
rs137921340 Chr.8: 130821416-130821416 on Build GRCh38
rs1027143550 Chr.8: 130821319-130821319 on Build GRCh38
rs201691369 Chr.8: 130821431-130821431 on Build GRCh38
rs772789667 Chr.8: 130821364-130821364 on Build GRCh38
rs145363107 Chr.8: 130821408-130821408 on Build GRCh38
rs200821614 Chr.8: 130821332-130821332 on Build GRCh38
rs866416086 Chr.8: 130821287-130821287 on Build GRCh38
rs753297735 Chr.8: 130821395-130821395 on Build GRCh38
rs746395435 Chr.8: 130821313-130821313 on Build GRCh38
rs781434048 Chr.8: 130821432-130821432 on Build GRCh38
rs199832182 Chr.8: 130821430-130821430 on Build GRCh38
rs921229749 Chr.8: 130821390-130821390 on Build GRCh38
rs908044977 Chr.8: 130821454-130821454 on Build GRCh38
rs745875308 Chr.8: 130821458-130821458 on Build GRCh38
rs983300002 Chr.8: 130821328-130821328 on Build GRCh38
rs749781288 Chr.8: 130821466-130821466 on Build GRCh38
rs780763478 Chr.8: 130821460-130821460 on Build GRCh38
rs534408380 Chr.8: 130821386-130821386 on Build GRCh38
rs189616227 Chr.8: 130821480-130821480 on Build GRCh38
rs866344510 Chr.8: 130821389-130821389 on Build GRCh38
rs749159194 Chr.8: 130821270-130821270 on Build GRCh38
rs776808750 Chr.8: 130821312-130821312 on Build GRCh38
rs768943774 Chr.8: 130821470-130821470 on Build GRCh38
rs980088169 Chr.8: 130821338-130821338 on Build GRCh38
rs370039220 Chr.8: 130821301-130821301 on Build GRCh38
rs760345728 Chr.8: 130821377-130821377 on Build GRCh38
rs765096807 Chr.8: 130821414-130821414 on Build GRCh38
rs765905342 Chr.8: 130821380-130821380 on Build GRCh38
rs545213357 Chr.8: 130821473-130821473 on Build GRCh38
rs373313882 Chr.8: 130821327-130821327 on Build GRCh38
rs1008502850 Chr.8: 130821299-130821299 on Build GRCh38
rs372263958 Chr.8: 130821310-130821310 on Build GRCh38
rs377504346 Chr.8: 130821351-130821351 on Build GRCh38
rs1057456527 Chr.8: 130821424-130821424 on Build GRCh38
rs370127292 Chr.8: 130821406-130821406 on Build GRCh38
rs1033285607 Chr.8: 130821273-130821273 on Build GRCh38

View Full Product Details