Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49276550-49276805 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf62
Entrez Gene ID
375341
Gene Name
chromosome 3 open reading frame 62
Gene Aliases
-
UniGene
Hs.403828
Gene Chromosome Location
Chr.3: 49268597-49277075 on Build GRCh38
Transcripts
NM_198562.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5962528 c.265G>A p.E89K Chr.3: 49276608-49276608 on Build GRCh38
1309258 c.97G>A p.E33K Chr.3: 49276776-49276776 on Build GRCh38
1423927 c.275C>T p.A92V Chr.3: 49276598-49276598 on Build GRCh38
5864799 c.238G>C p.A80P Chr.3: 49276635-49276635 on Build GRCh38
7358551 c.196A>G p.R66G Chr.3: 49276677-49276677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780482699 Chr.3: 49276745-49276745 on Build GRCh38
rs537882513 Chr.3: 49276704-49276706 on Build GRCh38
rs748190744 Chr.3: 49276686-49276686 on Build GRCh38
rs113941560 Chr.3: 49276706-49276706 on Build GRCh38
rs752204866 Chr.3: 49276635-49276635 on Build GRCh38
rs765399300 Chr.3: 49276596-49276596 on Build GRCh38
rs756790673 Chr.3: 49276742-49276742 on Build GRCh38
rs763891950 Chr.3: 49276663-49276664 on Build GRCh38
rs762705614 Chr.3: 49276603-49276603 on Build GRCh38
rs773022912 Chr.3: 49276598-49276598 on Build GRCh38
rs61749312 Chr.3: 49276695-49276695 on Build GRCh38
rs752294916 Chr.3: 49276726-49276726 on Build GRCh38
rs767316320 Chr.3: 49276725-49276725 on Build GRCh38
rs767066270 Chr.3: 49276618-49276618 on Build GRCh38
rs770044127 Chr.3: 49276687-49276687 on Build GRCh38
rs375872034 Chr.3: 49276653-49276653 on Build GRCh38
rs766766358 Chr.3: 49276637-49276637 on Build GRCh38
rs765116060 Chr.3: 49276761-49276761 on Build GRCh38
rs753561198 Chr.3: 49276695-49276696 on Build GRCh38
rs753533071 Chr.3: 49276738-49276738 on Build GRCh38
rs1020358122 Chr.3: 49276720-49276720 on Build GRCh38
rs750140796 Chr.3: 49276768-49276768 on Build GRCh38
rs758774110 Chr.3: 49276660-49276660 on Build GRCh38
rs751824683 Chr.3: 49276760-49276760 on Build GRCh38
rs780490292 Chr.3: 49276674-49276674 on Build GRCh38
rs763894323 Chr.3: 49276734-49276734 on Build GRCh38
rs754804103 Chr.3: 49276616-49276616 on Build GRCh38
rs1001466873 Chr.3: 49276652-49276652 on Build GRCh38
rs760534786 Chr.3: 49276636-49276637 on Build GRCh38
rs199624732 Chr.3: 49276783-49276783 on Build GRCh38
rs755862872 Chr.3: 49276641-49276641 on Build GRCh38
rs932467535 Chr.3: 49276623-49276623 on Build GRCh38
rs967903493 Chr.3: 49276736-49276736 on Build GRCh38
rs557579251 Chr.3: 49276684-49276684 on Build GRCh38
rs755543376 Chr.3: 49276763-49276763 on Build GRCh38
rs762039436 Chr.3: 49276583-49276583 on Build GRCh38
rs866948396 Chr.3: 49276715-49276715 on Build GRCh38
rs371684697 Chr.3: 49276599-49276599 on Build GRCh38
rs878948441 Chr.3: 49276722-49276722 on Build GRCh38
rs760235834 Chr.3: 49276730-49276730 on Build GRCh38
rs766278125 Chr.3: 49276609-49276609 on Build GRCh38
rs751502908 Chr.3: 49276615-49276615 on Build GRCh38
rs61749309 Chr.3: 49276677-49276677 on Build GRCh38
rs781641954 Chr.3: 49276779-49276779 on Build GRCh38
rs200997728 Chr.3: 49276781-49276781 on Build GRCh38
rs1045428487 Chr.3: 49276643-49276643 on Build GRCh38
rs17855113 Chr.3: 49276663-49276663 on Build GRCh38
rs759538759 Chr.3: 49276707-49276707 on Build GRCh38
rs766556144 Chr.3: 49276763-49276766 on Build GRCh38
rs774667709 Chr.3: 49276649-49276649 on Build GRCh38
rs921362347 Chr.3: 49276621-49276621 on Build GRCh38

View Full Product Details