Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87984165-87984430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BANP
Entrez Gene ID
54971
Gene Name
BTG3 associated nuclear protein
Gene Aliases
BEND1, SMAR1, SMARBP1
UniGene
Hs.461705
Gene Chromosome Location
Chr.16: 87949216-88077318 on Build GRCh38
Transcripts
NM_001173539.1 NM_001173540.1 NM_001173541.1 NM_001173542.1 NM_001173543.1 NM_017869.3 NM_079837.2 XM_005256008.3 XM_005256009.3 XM_011523170.2 XM_011523172.2 XM_011523174.2 XM_011523175.2 XM_011523176.2 XM_011523177.2 XM_011523178.2 XM_011523179.2 XM_011523180.2 XM_011523181.2 XM_011523182.1 XM_011523183.2 XM_011523184.2 XM_011523186.2 XM_017023378.1 XM_017023379.1 XM_017023380.1 XM_017023381.1 XM_017023383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4944887 c.338+1G>A p.? Chr.16: 87984260-87984260 on Build GRCh38
4594780 c.324C>T p.V108V Chr.16: 87984203-87984203 on Build GRCh38
7293254 c.269G>A p.S90N Chr.16: 87984190-87984190 on Build GRCh38
4594779 c.282C>T p.V94V Chr.16: 87984203-87984203 on Build GRCh38
5734083 c.341C>T p.S114F Chr.16: 87984220-87984220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137943095 Chr.16: 87984309-87984309 on Build GRCh38
rs775701963 Chr.16: 87984276-87984276 on Build GRCh38
rs928228619 Chr.16: 87984284-87984284 on Build GRCh38
rs770193546 Chr.16: 87984270-87984270 on Build GRCh38
rs966714338 Chr.16: 87984396-87984396 on Build GRCh38
rs550337364 Chr.16: 87984280-87984280 on Build GRCh38
rs958131693 Chr.16: 87984268-87984268 on Build GRCh38
rs547536427 Chr.16: 87984195-87984195 on Build GRCh38
rs989501535 Chr.16: 87984385-87984385 on Build GRCh38
rs768787794 Chr.16: 87984281-87984281 on Build GRCh38
rs374004218 Chr.16: 87984227-87984227 on Build GRCh38
rs183689725 Chr.16: 87984368-87984368 on Build GRCh38
rs886735937 Chr.16: 87984316-87984316 on Build GRCh38
rs758937392 Chr.16: 87984215-87984215 on Build GRCh38
rs1055280315 Chr.16: 87984297-87984297 on Build GRCh38
rs774099586 Chr.16: 87984285-87984285 on Build GRCh38
rs540840182 Chr.16: 87984334-87984338 on Build GRCh38
rs753345817 Chr.16: 87984208-87984208 on Build GRCh38
rs901088258 Chr.16: 87984331-87984331 on Build GRCh38
rs140242915 Chr.16: 87984191-87984191 on Build GRCh38
rs755803797 Chr.16: 87984206-87984206 on Build GRCh38
rs539595280 Chr.16: 87984291-87984291 on Build GRCh38
rs763999087 Chr.16: 87984190-87984190 on Build GRCh38
rs146413083 Chr.16: 87984240-87984240 on Build GRCh38
rs746061067 Chr.16: 87984269-87984269 on Build GRCh38
rs975265838 Chr.16: 87984243-87984243 on Build GRCh38
rs147212049 Chr.16: 87984344-87984344 on Build GRCh38
rs761449928 Chr.16: 87984194-87984194 on Build GRCh38
rs775757102 Chr.16: 87984188-87984188 on Build GRCh38
rs748191141 Chr.16: 87984281-87984281 on Build GRCh38
rs140824678 Chr.16: 87984242-87984242 on Build GRCh38
rs767259576 Chr.16: 87984308-87984308 on Build GRCh38
rs1019983866 Chr.16: 87984328-87984328 on Build GRCh38
rs765851784 Chr.16: 87984207-87984207 on Build GRCh38
rs35251544 Chr.16: 87984364-87984364 on Build GRCh38
rs576662808 Chr.16: 87984310-87984310 on Build GRCh38
rs560236562 Chr.16: 87984362-87984362 on Build GRCh38
rs1028867837 Chr.16: 87984247-87984247 on Build GRCh38
rs376119191 Chr.16: 87984235-87984235 on Build GRCh38
rs574310287 Chr.16: 87984332-87984332 on Build GRCh38
rs3815820 Chr.16: 87984203-87984203 on Build GRCh38
rs143709693 Chr.16: 87984321-87984321 on Build GRCh38
rs757511257 Chr.16: 87984241-87984241 on Build GRCh38
rs867711835 Chr.16: 87984397-87984397 on Build GRCh38
rs181040972 Chr.16: 87984313-87984313 on Build GRCh38

View Full Product Details