Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96115620-96115881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRA2B
Entrez Gene ID
151
Gene Name
adrenoceptor alpha 2B
Gene Aliases
ADRA2L1, ADRA2RL1, ADRARL1, ALPHA2BAR, FAME2, alpha-2BAR
UniGene
Hs.247686
Gene Chromosome Location
Chr.2: 96112875-96116245 on Build GRCh38
Transcripts
NM_000682.6

SNP Detail

SNP ID Chromosome Location
rs369790085 Chr.2: 96115810-96115810 on Build GRCh38
rs79421642 Chr.2: 96115746-96115746 on Build GRCh38
rs375435216 Chr.2: 96115788-96115788 on Build GRCh38
rs375034155 Chr.2: 96115727-96115727 on Build GRCh38
rs755785109 Chr.2: 96115803-96115803 on Build GRCh38
rs547122702 Chr.2: 96115775-96115775 on Build GRCh38
rs778813149 Chr.2: 96115757-96115757 on Build GRCh38
rs766562145 Chr.2: 96115674-96115674 on Build GRCh38
rs893501137 Chr.2: 96115820-96115820 on Build GRCh38
rs368204339 Chr.2: 96115857-96115857 on Build GRCh38
rs747882206 Chr.2: 96115767-96115767 on Build GRCh38
rs756992097 Chr.2: 96115713-96115713 on Build GRCh38
rs369353304 Chr.2: 96115673-96115673 on Build GRCh38
rs759694982 Chr.2: 96115769-96115769 on Build GRCh38
rs779659371 Chr.2: 96115733-96115733 on Build GRCh38
rs529425780 Chr.2: 96115659-96115659 on Build GRCh38
rs1012841819 Chr.2: 96115728-96115728 on Build GRCh38
rs556152097 Chr.2: 96115856-96115856 on Build GRCh38
rs776748854 Chr.2: 96115675-96115675 on Build GRCh38
rs950253015 Chr.2: 96115787-96115787 on Build GRCh38
rs769983087 Chr.2: 96115823-96115823 on Build GRCh38
rs760448500 Chr.2: 96115859-96115859 on Build GRCh38
rs749124572 Chr.2: 96115667-96115667 on Build GRCh38
rs769895462 Chr.2: 96115772-96115772 on Build GRCh38
rs763331228 Chr.2: 96115693-96115693 on Build GRCh38
rs976423590 Chr.2: 96115831-96115831 on Build GRCh38
rs771208116 Chr.2: 96115768-96115768 on Build GRCh38
rs775577204 Chr.2: 96115660-96115660 on Build GRCh38
rs570328500 Chr.2: 96115798-96115798 on Build GRCh38
rs757603778 Chr.2: 96115783-96115793 on Build GRCh38
rs780992353 Chr.2: 96115721-96115721 on Build GRCh38
rs759607097 Chr.2: 96115677-96115677 on Build GRCh38
rs13306147 Chr.2: 96115712-96115712 on Build GRCh38
rs370055953 Chr.2: 96115844-96115844 on Build GRCh38
rs752032229 Chr.2: 96115796-96115796 on Build GRCh38
rs775403096 Chr.2: 96115773-96115773 on Build GRCh38
rs202241079 Chr.2: 96115837-96115837 on Build GRCh38
rs777494477 Chr.2: 96115821-96115821 on Build GRCh38
rs748051200 Chr.2: 96115816-96115816 on Build GRCh38
rs768443018 Chr.2: 96115752-96115752 on Build GRCh38
rs771866132 Chr.2: 96115818-96115818 on Build GRCh38
rs753262027 Chr.2: 96115804-96115804 on Build GRCh38
rs753258709 Chr.2: 96115687-96115687 on Build GRCh38
rs372554591 Chr.2: 96115811-96115811 on Build GRCh38
rs372831901 Chr.2: 96115697-96115697 on Build GRCh38
rs765233575 Chr.2: 96115681-96115681 on Build GRCh38
rs768515922 Chr.2: 96115671-96115671 on Build GRCh38
rs539438565 Chr.2: 96115822-96115822 on Build GRCh38
rs549167238 Chr.2: 96115672-96115672 on Build GRCh38
rs768718519 Chr.2: 96115838-96115838 on Build GRCh38
rs564024418 Chr.2: 96115641-96115641 on Build GRCh38
rs759921636 Chr.2: 96115669-96115669 on Build GRCh38
rs750125417 Chr.2: 96115802-96115802 on Build GRCh38
rs1046536596 Chr.2: 96115797-96115797 on Build GRCh38

View Full Product Details