Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 90916170-90916399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD1
Entrez Gene ID
27063
Gene Name
ankyrin repeat domain 1
Gene Aliases
ALRP, C-193, CARP, CVARP, MCARP, bA320F15.2
UniGene
Hs.448589
Gene Chromosome Location
Chr.10: 90912100-90921275 on Build GRCh38
Transcripts
NM_014391.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4554597 c.625G>A p.G209R Chr.10: 90916197-90916197 on Build GRCh38
4818943 c.646G>A p.D216N Chr.10: 90916176-90916176 on Build GRCh38
5589655 c.590G>A p.G197E Chr.10: 90916232-90916232 on Build GRCh38
6537076 c.613T>C p.L205L Chr.10: 90916209-90916209 on Build GRCh38
4777282 c.626G>C p.G209A Chr.10: 90916196-90916196 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs868668600 Chr.10: 90916224-90916224 on Build GRCh38
rs769305030 Chr.10: 90916239-90916239 on Build GRCh38
rs1003651682 Chr.10: 90916290-90916290 on Build GRCh38
rs754771221 Chr.10: 90916197-90916197 on Build GRCh38
rs1037418556 Chr.10: 90916366-90916366 on Build GRCh38
rs749531136 Chr.10: 90916274-90916275 on Build GRCh38
rs775996412 Chr.10: 90916288-90916288 on Build GRCh38
rs797017029 Chr.10: 90916222-90916223 on Build GRCh38
rs4933200 Chr.10: 90916370-90916370 on Build GRCh38
rs915573585 Chr.10: 90916364-90916364 on Build GRCh38
rs992192584 Chr.10: 90916325-90916325 on Build GRCh38
rs375020388 Chr.10: 90916296-90916296 on Build GRCh38
rs775666061 Chr.10: 90916269-90916270 on Build GRCh38
rs557044291 Chr.10: 90916356-90916356 on Build GRCh38
rs538585155 Chr.10: 90916225-90916225 on Build GRCh38
rs397517251 Chr.10: 90916262-90916262 on Build GRCh38
rs769158987 Chr.10: 90916297-90916297 on Build GRCh38
rs867949780 Chr.10: 90916233-90916233 on Build GRCh38
rs1018915821 Chr.10: 90916284-90916284 on Build GRCh38
rs869025363 Chr.10: 90916210-90916210 on Build GRCh38
rs768491548 Chr.10: 90916313-90916313 on Build GRCh38
rs769897907 Chr.10: 90916253-90916253 on Build GRCh38
rs1004430284 Chr.10: 90916223-90916223 on Build GRCh38
rs781049118 Chr.10: 90916219-90916219 on Build GRCh38
rs777185169 Chr.10: 90916252-90916252 on Build GRCh38
rs794728974 Chr.10: 90916269-90916269 on Build GRCh38
rs941687322 Chr.10: 90916365-90916365 on Build GRCh38
rs770544020 Chr.10: 90916286-90916286 on Build GRCh38
rs748104942 Chr.10: 90916238-90916238 on Build GRCh38

View Full Product Details