Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 11545943-11546214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLK
Entrez Gene ID
640
Gene Name
BLK proto-oncogene, Src family tyrosine kinase
Gene Aliases
MODY11
UniGene
Hs.146591
Gene Chromosome Location
Chr.8: 11493991-11564599 on Build GRCh38
Transcripts
NM_001715.2 XM_011543824.1 XM_011543825.2 XM_011543827.1 XM_011543828.2 XM_011543829.2 XM_017013730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3884971 c.152C>T p.P51L Chr.8: 11546080-11546080 on Build GRCh38
6611757 c.135C>T p.N45N Chr.8: 11546063-11546063 on Build GRCh38
231148 c.145C>T p.P49S Chr.8: 11546073-11546073 on Build GRCh38
3698794 c.166C>A p.L56M Chr.8: 11546094-11546094 on Build GRCh38
748583 c.153G>T p.P51P Chr.8: 11546081-11546081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146140570 Chr.8: 11546074-11546074 on Build GRCh38
rs1051351361 Chr.8: 11546033-11546033 on Build GRCh38
rs776904190 Chr.8: 11546126-11546126 on Build GRCh38
rs796310144 Chr.8: 11546176-11546176 on Build GRCh38
rs752656205 Chr.8: 11546131-11546131 on Build GRCh38
rs776158048 Chr.8: 11546020-11546020 on Build GRCh38
rs745428886 Chr.8: 11546084-11546084 on Build GRCh38
rs900563995 Chr.8: 11546170-11546170 on Build GRCh38
rs1039155851 Chr.8: 11546159-11546159 on Build GRCh38
rs372940637 Chr.8: 11546034-11546034 on Build GRCh38
rs779948480 Chr.8: 11546001-11546001 on Build GRCh38
rs202053568 Chr.8: 11546092-11546092 on Build GRCh38
rs761075080 Chr.8: 11546109-11546109 on Build GRCh38
rs773488674 Chr.8: 11546103-11546103 on Build GRCh38
rs968165628 Chr.8: 11546146-11546146 on Build GRCh38
rs753844347 Chr.8: 11546004-11546004 on Build GRCh38
rs777409945 Chr.8: 11546011-11546011 on Build GRCh38
rs373519246 Chr.8: 11546012-11546012 on Build GRCh38
rs202045056 Chr.8: 11546067-11546067 on Build GRCh38
rs534417637 Chr.8: 11545971-11545971 on Build GRCh38
rs772595681 Chr.8: 11546098-11546098 on Build GRCh38
rs147622122 Chr.8: 11546023-11546023 on Build GRCh38
rs373261122 Chr.8: 11546140-11546140 on Build GRCh38
rs964607017 Chr.8: 11545974-11545974 on Build GRCh38
rs377676972 Chr.8: 11546085-11546085 on Build GRCh38
rs765904855 Chr.8: 11545990-11545990 on Build GRCh38
rs775010605 Chr.8: 11546086-11546086 on Build GRCh38
rs377480038 Chr.8: 11546016-11546016 on Build GRCh38
rs762337317 Chr.8: 11546030-11546030 on Build GRCh38
rs765245967 Chr.8: 11546129-11546129 on Build GRCh38
rs764346696 Chr.8: 11546123-11546123 on Build GRCh38
rs374755485 Chr.8: 11546148-11546148 on Build GRCh38
rs377568491 Chr.8: 11546057-11546057 on Build GRCh38
rs769336709 Chr.8: 11546027-11546027 on Build GRCh38
rs375173243 Chr.8: 11546082-11546082 on Build GRCh38
rs571333916 Chr.8: 11546054-11546054 on Build GRCh38
rs778356577 Chr.8: 11546185-11546185 on Build GRCh38
rs757062096 Chr.8: 11546145-11546145 on Build GRCh38
rs370556587 Chr.8: 11546066-11546066 on Build GRCh38
rs771847711 Chr.8: 11546009-11546009 on Build GRCh38
rs750695701 Chr.8: 11546047-11546047 on Build GRCh38
rs747992550 Chr.8: 11546008-11546008 on Build GRCh38
rs745460951 Chr.8: 11546151-11546151 on Build GRCh38
rs997753667 Chr.8: 11545979-11545979 on Build GRCh38
rs140188373 Chr.8: 11546080-11546080 on Build GRCh38
rs1035007550 Chr.8: 11546107-11546107 on Build GRCh38
rs751360261 Chr.8: 11546143-11546143 on Build GRCh38
rs2244938 Chr.8: 11546134-11546134 on Build GRCh38
rs754908449 Chr.8: 11546007-11546007 on Build GRCh38
rs35339715 Chr.8: 11546071-11546071 on Build GRCh38
rs746710451 Chr.8: 11546081-11546081 on Build GRCh38
rs759537061 Chr.8: 11546128-11546128 on Build GRCh38
rs371523216 Chr.8: 11545996-11545996 on Build GRCh38
rs774986468 Chr.8: 11546029-11546029 on Build GRCh38
rs1030277156 Chr.8: 11545987-11545987 on Build GRCh38
rs754499170 Chr.8: 11546180-11546180 on Build GRCh38
rs35298605 Chr.8: 11546093-11546093 on Build GRCh38
rs766564806 Chr.8: 11546113-11546113 on Build GRCh38
rs924149175 Chr.8: 11546166-11546166 on Build GRCh38

View Full Product Details