Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99316121-99316378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1B
Entrez Gene ID
125
Gene Name
alcohol dehydrogenase 1B (class I), beta polypeptide
Gene Aliases
ADH2, HEL-S-117
UniGene
Hs.4
Gene Chromosome Location
Chr.4: 99306387-99321442 on Build GRCh38
Transcripts
NM_000668.5 NM_001286650.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3598575 c.262G>A p.D88N Chr.4: 99316300-99316300 on Build GRCh38
3598574 c.274C>T p.P92S Chr.4: 99316288-99316288 on Build GRCh38
5356188 c.303C>T p.C101C Chr.4: 99316259-99316259 on Build GRCh38
4120943 c.336C>T p.C112C Chr.4: 99316226-99316226 on Build GRCh38
7329969 c.300A>G p.K100K Chr.4: 99316262-99316262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55806158 Chr.4: 99316192-99316193 on Build GRCh38
rs575310578 Chr.4: 99316152-99316152 on Build GRCh38
rs771688874 Chr.4: 99316157-99316157 on Build GRCh38
rs755072205 Chr.4: 99316345-99316345 on Build GRCh38
rs201292240 Chr.4: 99316175-99316175 on Build GRCh38
rs914113960 Chr.4: 99316246-99316246 on Build GRCh38
rs757632781 Chr.4: 99316217-99316217 on Build GRCh38
rs767886585 Chr.4: 99316287-99316287 on Build GRCh38
rs760362364 Chr.4: 99316269-99316269 on Build GRCh38
rs752089296 Chr.4: 99316283-99316283 on Build GRCh38
rs971026459 Chr.4: 99316250-99316250 on Build GRCh38
rs148622183 Chr.4: 99316242-99316242 on Build GRCh38
rs200395036 Chr.4: 99316216-99316216 on Build GRCh38
rs540222176 Chr.4: 99316306-99316306 on Build GRCh38
rs372958183 Chr.4: 99316253-99316253 on Build GRCh38
rs924362380 Chr.4: 99316326-99316326 on Build GRCh38
rs775840233 Chr.4: 99316255-99316256 on Build GRCh38
rs765919126 Chr.4: 99316186-99316186 on Build GRCh38
rs749350311 Chr.4: 99316241-99316241 on Build GRCh38
rs748051173 Chr.4: 99316350-99316350 on Build GRCh38
rs772986488 Chr.4: 99316167-99316167 on Build GRCh38
rs376805756 Chr.4: 99316227-99316227 on Build GRCh38
rs560773951 Chr.4: 99316236-99316236 on Build GRCh38
rs749342166 Chr.4: 99316238-99316238 on Build GRCh38
rs370001112 Chr.4: 99316332-99316332 on Build GRCh38
rs756380510 Chr.4: 99316322-99316322 on Build GRCh38
rs369184871 Chr.4: 99316243-99316243 on Build GRCh38
rs753987123 Chr.4: 99316340-99316340 on Build GRCh38
rs4147539 Chr.4: 99316323-99316325 on Build GRCh38
rs952025294 Chr.4: 99316219-99316219 on Build GRCh38
rs74732879 Chr.4: 99316214-99316214 on Build GRCh38
rs7435646 Chr.4: 99316335-99316335 on Build GRCh38
rs770585785 Chr.4: 99316270-99316270 on Build GRCh38
rs945604428 Chr.4: 99316273-99316273 on Build GRCh38
rs528239306 Chr.4: 99316281-99316281 on Build GRCh38
rs375685735 Chr.4: 99316272-99316272 on Build GRCh38
rs993694773 Chr.4: 99316144-99316144 on Build GRCh38
rs764637094 Chr.4: 99316212-99316212 on Build GRCh38
rs778728860 Chr.4: 99316346-99316346 on Build GRCh38
rs773002927 Chr.4: 99316260-99316260 on Build GRCh38
rs146657145 Chr.4: 99316286-99316286 on Build GRCh38
rs763502168 Chr.4: 99316206-99316206 on Build GRCh38
rs372446680 Chr.4: 99316328-99316330 on Build GRCh38
rs753286056 Chr.4: 99316198-99316198 on Build GRCh38
rs142065383 Chr.4: 99316276-99316276 on Build GRCh38
rs771842423 Chr.4: 99316255-99316255 on Build GRCh38

View Full Product Details