Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 12816490-12816729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAND2
Entrez Gene ID
23066
Gene Name
cullin associated and neddylation dissociated 2 (putative)
Gene Aliases
TIP120B, Tp120b
UniGene
Hs.343664
Gene Chromosome Location
Chr.3: 12796654-12834818 on Build GRCh38
Transcripts
NM_001162499.1 NM_012298.2 XM_011533503.2 XM_011533504.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3974146 c.1571C>T p.P524L Chr.3: 12816503-12816503 on Build GRCh38
7297574 c.1449G>A p.A483A Chr.3: 12816660-12816660 on Build GRCh38
4618291 c.1331T>C p.I444T Chr.3: 12816542-12816542 on Build GRCh38
3914862 c.1285C>T p.L429F Chr.3: 12816496-12816496 on Build GRCh38
3974147 c.1292C>T p.P431L Chr.3: 12816503-12816503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753838369 Chr.3: 12816580-12816580 on Build GRCh38
rs755380982 Chr.3: 12816681-12816681 on Build GRCh38
rs369765807 Chr.3: 12816593-12816593 on Build GRCh38
rs747742827 Chr.3: 12816513-12816513 on Build GRCh38
rs377274058 Chr.3: 12816581-12816581 on Build GRCh38
rs745389970 Chr.3: 12816702-12816702 on Build GRCh38
rs375865585 Chr.3: 12816662-12816662 on Build GRCh38
rs370736670 Chr.3: 12816549-12816549 on Build GRCh38
rs767358207 Chr.3: 12816546-12816546 on Build GRCh38
rs1044429842 Chr.3: 12816553-12816553 on Build GRCh38
rs756860213 Chr.3: 12816699-12816699 on Build GRCh38
rs145281444 Chr.3: 12816659-12816659 on Build GRCh38
rs372784797 Chr.3: 12816661-12816661 on Build GRCh38
rs373922459 Chr.3: 12816602-12816602 on Build GRCh38
rs780179047 Chr.3: 12816710-12816710 on Build GRCh38
rs760936753 Chr.3: 12816550-12816550 on Build GRCh38
rs774390761 Chr.3: 12816537-12816537 on Build GRCh38
rs200083326 Chr.3: 12816627-12816627 on Build GRCh38
rs751901449 Chr.3: 12816679-12816679 on Build GRCh38
rs201969409 Chr.3: 12816629-12816629 on Build GRCh38
rs763813570 Chr.3: 12816538-12816538 on Build GRCh38
rs778444982 Chr.3: 12816701-12816701 on Build GRCh38
rs3732675 Chr.3: 12816529-12816529 on Build GRCh38
rs769441536 Chr.3: 12816515-12816515 on Build GRCh38
rs763411636 Chr.3: 12816668-12816668 on Build GRCh38
rs764432053 Chr.3: 12816559-12816559 on Build GRCh38
rs777291094 Chr.3: 12816686-12816686 on Build GRCh38
rs745749653 Chr.3: 12816633-12816633 on Build GRCh38
rs766897566 Chr.3: 12816675-12816675 on Build GRCh38
rs772672304 Chr.3: 12816522-12816522 on Build GRCh38
rs201492931 Chr.3: 12816647-12816647 on Build GRCh38
rs113368650 Chr.3: 12816594-12816594 on Build GRCh38
rs765658921 Chr.3: 12816590-12816590 on Build GRCh38
rs777455787 Chr.3: 12816620-12816620 on Build GRCh38
rs974805981 Chr.3: 12816665-12816665 on Build GRCh38
rs923697036 Chr.3: 12816709-12816709 on Build GRCh38
rs775374995 Chr.3: 12816648-12816648 on Build GRCh38
rs753469137 Chr.3: 12816690-12816690 on Build GRCh38
rs748719033 Chr.3: 12816621-12816621 on Build GRCh38
rs759234744 Chr.3: 12816545-12816545 on Build GRCh38
rs760593457 Chr.3: 12816653-12816653 on Build GRCh38
rs772112640 Chr.3: 12816704-12816704 on Build GRCh38
rs574738317 Chr.3: 12816607-12816607 on Build GRCh38
rs750475682 Chr.3: 12816667-12816667 on Build GRCh38
rs780331728 Chr.3: 12816596-12816596 on Build GRCh38
rs755717713 Chr.3: 12816611-12816611 on Build GRCh38

View Full Product Details