Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241666798-241667051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG4B
Entrez Gene ID
23192
Gene Name
autophagy related 4B cysteine peptidase
Gene Aliases
APG4B, AUTL1
UniGene
Hs.283610
Gene Chromosome Location
Chr.2: 241637612-241673857 on Build GRCh38
Transcripts
NM_013325.4 NM_178326.2 XM_005246992.3 XM_005246993.2 XM_005246995.3 XM_005246996.2 XM_017003637.1 XM_017003638.1 XM_017003639.1 XM_017003640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
477188 c.726G>T p.T242T Chr.2: 241666832-241666832 on Build GRCh38
6999452 c.676G>A p.V226M Chr.2: 241666824-241666824 on Build GRCh38
6999451 c.718G>A p.V240M Chr.2: 241666824-241666824 on Build GRCh38
6999450 c.718G>A p.V240M Chr.2: 241666824-241666824 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs144028955 Chr.2: 241666858-241666858 on Build GRCh38
rs1028084699 Chr.2: 241666976-241666976 on Build GRCh38
rs766529063 Chr.2: 241666851-241666851 on Build GRCh38
rs369350394 Chr.2: 241666854-241666854 on Build GRCh38
rs916041271 Chr.2: 241667026-241667026 on Build GRCh38
rs373542750 Chr.2: 241666877-241666877 on Build GRCh38
rs963530634 Chr.2: 241667007-241667007 on Build GRCh38
rs867543451 Chr.2: 241666944-241666944 on Build GRCh38
rs550817093 Chr.2: 241666921-241666921 on Build GRCh38
rs201831892 Chr.2: 241666848-241666848 on Build GRCh38
rs535532298 Chr.2: 241666999-241666999 on Build GRCh38
rs1017585080 Chr.2: 241667005-241667005 on Build GRCh38
rs1018954842 Chr.2: 241666989-241666989 on Build GRCh38
rs904957554 Chr.2: 241666954-241666954 on Build GRCh38
rs569053147 Chr.2: 241666925-241666925 on Build GRCh38
rs990302503 Chr.2: 241666883-241666883 on Build GRCh38
rs745993603 Chr.2: 241666915-241666915 on Build GRCh38
rs771019497 Chr.2: 241666855-241666855 on Build GRCh38
rs762166176 Chr.2: 241666831-241666831 on Build GRCh38
rs183235226 Chr.2: 241666916-241666916 on Build GRCh38
rs929932298 Chr.2: 241666920-241666920 on Build GRCh38
rs949072220 Chr.2: 241666942-241666942 on Build GRCh38
rs753832972 Chr.2: 241666853-241666853 on Build GRCh38
rs201063196 Chr.2: 241666850-241666850 on Build GRCh38
rs758109088 Chr.2: 241666867-241666867 on Build GRCh38
rs1001013676 Chr.2: 241666957-241666957 on Build GRCh38
rs915676479 Chr.2: 241666912-241666912 on Build GRCh38
rs973876037 Chr.2: 241667024-241667024 on Build GRCh38
rs547476442 Chr.2: 241666938-241666938 on Build GRCh38
rs561638837 Chr.2: 241666913-241666913 on Build GRCh38
rs370063366 Chr.2: 241666875-241666875 on Build GRCh38
rs534715760 Chr.2: 241666947-241666947 on Build GRCh38
rs533301541 Chr.2: 241666933-241666933 on Build GRCh38
rs575560117 Chr.2: 241666824-241666824 on Build GRCh38
rs568481725 Chr.2: 241666973-241666973 on Build GRCh38
rs746622594 Chr.2: 241666874-241666874 on Build GRCh38
rs1008017100 Chr.2: 241666981-241666981 on Build GRCh38
rs1030604315 Chr.2: 241666977-241666977 on Build GRCh38
rs376762163 Chr.2: 241666871-241666871 on Build GRCh38
rs966479461 Chr.2: 241666991-241666991 on Build GRCh38
rs553296303 Chr.2: 241666959-241666959 on Build GRCh38
rs570874636 Chr.2: 241666946-241666946 on Build GRCh38
rs557164953 Chr.2: 241667008-241667008 on Build GRCh38
rs780732606 Chr.2: 241666884-241666885 on Build GRCh38
rs551695332 Chr.2: 241666939-241666939 on Build GRCh38
rs780611694 Chr.2: 241666878-241666878 on Build GRCh38
rs957010451 Chr.2: 241666832-241666832 on Build GRCh38
rs954920321 Chr.2: 241666998-241666998 on Build GRCh38
rs892611565 Chr.2: 241666965-241666965 on Build GRCh38
rs376746434 Chr.2: 241666842-241666842 on Build GRCh38
rs540179251 Chr.2: 241666908-241666908 on Build GRCh38

View Full Product Details