Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 59116545-59116806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB2
Entrez Gene ID
9133
Gene Name
cyclin B2
Gene Aliases
HsT17299
UniGene
Hs.194698
Gene Chromosome Location
Chr.15: 59105085-59125045 on Build GRCh38
Transcripts
NM_004701.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4450413 c.598-2A>G p.? Chr.15: 59116688-59116688 on Build GRCh38
6369917 c.605C>A p.P202Q Chr.15: 59116697-59116697 on Build GRCh38
700835 c.613C>T p.R205W Chr.15: 59116705-59116705 on Build GRCh38
470892 c.616A>T p.K206* Chr.15: 59116708-59116708 on Build GRCh38
3502797 c.611C>T p.S204F Chr.15: 59116703-59116703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055013442 Chr.15: 59116709-59116709 on Build GRCh38
rs146994389 Chr.15: 59116674-59116674 on Build GRCh38
rs764332149 Chr.15: 59116683-59116690 on Build GRCh38
rs754041645 Chr.15: 59116700-59116701 on Build GRCh38
rs574565044 Chr.15: 59116645-59116645 on Build GRCh38
rs28383540 Chr.15: 59116621-59116621 on Build GRCh38
rs778267846 Chr.15: 59116643-59116643 on Build GRCh38
rs756864076 Chr.15: 59116667-59116667 on Build GRCh38
rs767674929 Chr.15: 59116761-59116761 on Build GRCh38
rs768108919 Chr.15: 59116673-59116673 on Build GRCh38
rs750922699 Chr.15: 59116706-59116706 on Build GRCh38
rs933938759 Chr.15: 59116656-59116656 on Build GRCh38
rs548055725 Chr.15: 59116577-59116577 on Build GRCh38
rs774584434 Chr.15: 59116736-59116736 on Build GRCh38
rs921186021 Chr.15: 59116640-59116640 on Build GRCh38
rs28383542 Chr.15: 59116719-59116719 on Build GRCh38
rs759584289 Chr.15: 59116738-59116738 on Build GRCh38
rs941274347 Chr.15: 59116578-59116578 on Build GRCh38
rs201177056 Chr.15: 59116699-59116699 on Build GRCh38
rs28383541 Chr.15: 59116669-59116669 on Build GRCh38
rs756589572 Chr.15: 59116641-59116641 on Build GRCh38
rs556225520 Chr.15: 59116704-59116704 on Build GRCh38
rs776426387 Chr.15: 59116713-59116713 on Build GRCh38
rs369137460 Chr.15: 59116705-59116705 on Build GRCh38
rs150170652 Chr.15: 59116735-59116735 on Build GRCh38

View Full Product Details