Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 158671379-158671640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf46
Entrez Gene ID
201725
Gene Name
chromosome 4 open reading frame 46
Gene Aliases
RCDG1
UniGene
Hs.380920
Gene Chromosome Location
Chr.4: 158666675-158672255 on Build GRCh38
Transcripts
NM_001008393.3 NR_077234.1 NR_077235.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4330427 c.166G>C p.V56L Chr.4: 158671636-158671636 on Build GRCh38
1582618 c.178A>T p.I60F Chr.4: 158671624-158671624 on Build GRCh38
6616167 c.184G>T p.D62Y Chr.4: 158671618-158671618 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs528334389 Chr.4: 158671459-158671460 on Build GRCh38
rs925623672 Chr.4: 158671615-158671615 on Build GRCh38
rs1052225615 Chr.4: 158671490-158671490 on Build GRCh38
rs113661715 Chr.4: 158671486-158671486 on Build GRCh38
rs952213684 Chr.4: 158671606-158671606 on Build GRCh38
rs767081751 Chr.4: 158671607-158671608 on Build GRCh38
rs1027786226 Chr.4: 158671412-158671412 on Build GRCh38
rs935208355 Chr.4: 158671483-158671483 on Build GRCh38
rs976815488 Chr.4: 158671478-158671478 on Build GRCh38
rs910943277 Chr.4: 158671465-158671465 on Build GRCh38
rs746283617 Chr.4: 158671585-158671585 on Build GRCh38
rs373552594 Chr.4: 158671591-158671591 on Build GRCh38
rs112628838 Chr.4: 158671517-158671517 on Build GRCh38
rs144137757 Chr.4: 158671466-158671467 on Build GRCh38
rs536720153 Chr.4: 158671613-158671613 on Build GRCh38
rs200712046 Chr.4: 158671573-158671573 on Build GRCh38
rs983863733 Chr.4: 158671425-158671425 on Build GRCh38
rs1044884480 Chr.4: 158671498-158671498 on Build GRCh38
rs866100081 Chr.4: 158671442-158671442 on Build GRCh38
rs753342205 Chr.4: 158671620-158671620 on Build GRCh38
rs372933641 Chr.4: 158671587-158671587 on Build GRCh38
rs1027843440 Chr.4: 158671589-158671589 on Build GRCh38
rs376745396 Chr.4: 158671594-158671594 on Build GRCh38
rs111229175 Chr.4: 158671519-158671519 on Build GRCh38
rs779274304 Chr.4: 158671584-158671584 on Build GRCh38
rs993839778 Chr.4: 158671552-158671552 on Build GRCh38
rs759444014 Chr.4: 158671599-158671599 on Build GRCh38
rs185447209 Chr.4: 158671562-158671562 on Build GRCh38
rs1042779988 Chr.4: 158671429-158671429 on Build GRCh38
rs755726524 Chr.4: 158671611-158671611 on Build GRCh38
rs764558587 Chr.4: 158671441-158671441 on Build GRCh38
rs1016914886 Chr.4: 158671523-158671523 on Build GRCh38
rs758979858 Chr.4: 158671585-158671586 on Build GRCh38
rs954709203 Chr.4: 158671576-158671576 on Build GRCh38
rs369531296 Chr.4: 158671583-158671583 on Build GRCh38
rs767415410 Chr.4: 158671608-158671608 on Build GRCh38
rs905263807 Chr.4: 158671549-158671549 on Build GRCh38
rs567798319 Chr.4: 158671445-158671445 on Build GRCh38
rs386681193 Chr.4: 158671517-158671519 on Build GRCh38
rs942408532 Chr.4: 158671466-158671466 on Build GRCh38
rs190620255 Chr.4: 158671567-158671567 on Build GRCh38
rs775644595 Chr.4: 158671588-158671588 on Build GRCh38
rs528948936 Chr.4: 158671469-158671469 on Build GRCh38
rs548826949 Chr.4: 158671481-158671481 on Build GRCh38
rs572116257 Chr.4: 158671545-158671545 on Build GRCh38
rs550742771 Chr.4: 158671420-158671420 on Build GRCh38

View Full Product Details