Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202945024-202945257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIPOR1
Entrez Gene ID
51094
Gene Name
adiponectin receptor 1
Gene Aliases
ACDCR1, CGI-45, CGI45, PAQR1, TESBP1A
UniGene
Hs.5298
Gene Chromosome Location
Chr.1: 202940825-202958572 on Build GRCh38
Transcripts
NM_001290553.1 NM_001290557.1 NM_001290629.1 NM_015999.5 XM_017001407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7430278 c.449T>G p.F150C Chr.1: 202945151-202945151 on Build GRCh38
4660042 c.576C>T p.T192T Chr.1: 202945024-202945024 on Build GRCh38
6327415 c.513G>A p.V171V Chr.1: 202945087-202945087 on Build GRCh38
6324127 c.462G>T p.L154F Chr.1: 202945138-202945138 on Build GRCh38
1725359 c.457A>G p.I153V Chr.1: 202945143-202945143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761458096 Chr.1: 202945138-202945138 on Build GRCh38
rs760115326 Chr.1: 202945081-202945081 on Build GRCh38
rs770463342 Chr.1: 202945090-202945090 on Build GRCh38
rs1003880537 Chr.1: 202945083-202945083 on Build GRCh38
rs752673264 Chr.1: 202945180-202945180 on Build GRCh38
rs751230747 Chr.1: 202945129-202945129 on Build GRCh38
rs551807949 Chr.1: 202945045-202945045 on Build GRCh38
rs756017133 Chr.1: 202945165-202945165 on Build GRCh38
rs758506210 Chr.1: 202945194-202945194 on Build GRCh38
rs780018580 Chr.1: 202945167-202945167 on Build GRCh38
rs1027843095 Chr.1: 202945179-202945179 on Build GRCh38
rs149207948 Chr.1: 202945217-202945217 on Build GRCh38
rs775116917 Chr.1: 202945106-202945106 on Build GRCh38
rs201164064 Chr.1: 202945216-202945216 on Build GRCh38
rs767286210 Chr.1: 202945155-202945155 on Build GRCh38
rs146792118 Chr.1: 202945126-202945126 on Build GRCh38
rs1342387 Chr.1: 202945228-202945228 on Build GRCh38
rs769879784 Chr.1: 202945186-202945187 on Build GRCh38
rs777189732 Chr.1: 202945060-202945060 on Build GRCh38
rs1048097789 Chr.1: 202945203-202945203 on Build GRCh38
rs571874303 Chr.1: 202945048-202945048 on Build GRCh38
rs747609172 Chr.1: 202945187-202945187 on Build GRCh38
rs373089230 Chr.1: 202945117-202945117 on Build GRCh38
rs973430800 Chr.1: 202945153-202945153 on Build GRCh38
rs762544607 Chr.1: 202945118-202945118 on Build GRCh38
rs750288767 Chr.1: 202945164-202945164 on Build GRCh38

View Full Product Details