Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70774228-70774453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBATA
Entrez Gene ID
219793
Gene Name
thymus, brain and testes associated
Gene Aliases
C10orf27, SPATIAL
UniGene
Hs.386698
Gene Chromosome Location
Chr.10: 70771237-70785419 on Build GRCh38
Transcripts
NM_001318241.1 NM_001318242.1 NM_001318243.1 NM_152710.3 NR_134531.1 NR_134532.1 NR_134533.1 NR_134534.1 XM_017015845.1 XM_017015846.1 XM_017015847.1 XM_017015848.1 XM_017015849.1 XM_017015852.1 XM_017015853.1 XM_017015854.1 XM_017015855.1 XM_017015856.1 XM_017015857.1 XM_017015858.1 XM_017015859.1 XM_017015860.1 XM_017015861.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1702600 c.830C>T p.P277L Chr.10: 70774300-70774300 on Build GRCh38
6797084 c.886C>T p.P296S Chr.10: 70774244-70774244 on Build GRCh38
6869751 c.858C>T p.L286L Chr.10: 70774272-70774272 on Build GRCh38
1675324 c.778G>T p.D260Y Chr.10: 70774352-70774352 on Build GRCh38
3440141 c.895G>T p.E299* Chr.10: 70774235-70774235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983555148 Chr.10: 70774424-70774424 on Build GRCh38
rs897387630 Chr.10: 70774385-70774385 on Build GRCh38
rs944027096 Chr.10: 70774367-70774367 on Build GRCh38
rs1034081403 Chr.10: 70774434-70774434 on Build GRCh38
rs539891487 Chr.10: 70774326-70774326 on Build GRCh38
rs372705159 Chr.10: 70774372-70774372 on Build GRCh38
rs533241310 Chr.10: 70774346-70774346 on Build GRCh38
rs749163580 Chr.10: 70774287-70774287 on Build GRCh38
rs778931572 Chr.10: 70774390-70774390 on Build GRCh38
rs762764506 Chr.10: 70774329-70774329 on Build GRCh38
rs922188046 Chr.10: 70774296-70774296 on Build GRCh38
rs751554301 Chr.10: 70774396-70774396 on Build GRCh38
rs747162561 Chr.10: 70774301-70774301 on Build GRCh38
rs753705911 Chr.10: 70774373-70774373 on Build GRCh38
rs1012174147 Chr.10: 70774412-70774412 on Build GRCh38
rs1034229918 Chr.10: 70774393-70774393 on Build GRCh38
rs771513454 Chr.10: 70774338-70774338 on Build GRCh38
rs748457552 Chr.10: 70774324-70774324 on Build GRCh38
rs757119632 Chr.10: 70774382-70774382 on Build GRCh38
rs1024920391 Chr.10: 70774417-70774417 on Build GRCh38
rs773771976 Chr.10: 70774333-70774333 on Build GRCh38
rs768710623 Chr.10: 70774313-70774313 on Build GRCh38
rs970726708 Chr.10: 70774418-70774418 on Build GRCh38
rs566171083 Chr.10: 70774263-70774263 on Build GRCh38
rs150078967 Chr.10: 70774347-70774347 on Build GRCh38
rs537312540 Chr.10: 70774400-70774400 on Build GRCh38
rs373624773 Chr.10: 70774298-70774298 on Build GRCh38
rs139405249 Chr.10: 70774349-70774349 on Build GRCh38
rs942233577 Chr.10: 70774277-70774277 on Build GRCh38
rs200052414 Chr.10: 70774322-70774322 on Build GRCh38
rs888604055 Chr.10: 70774421-70774421 on Build GRCh38
rs143316854 Chr.10: 70774292-70774292 on Build GRCh38
rs907988632 Chr.10: 70774432-70774432 on Build GRCh38
rs763368087 Chr.10: 70774335-70774335 on Build GRCh38
rs555950093 Chr.10: 70774420-70774420 on Build GRCh38

View Full Product Details